Skip to Content
Merck
All Photos(1)

Key Documents

Y0001129

Ganciclovir

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Amino-1,9-dihydro-9-[[2-hydroxy-1-(hydroxymethyl)ethoxy]methyl]-6H-purin-6-one, 9-(1,3-Dihydroxy-2-propoxymethyl)guanine, DHPG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N5O4
CAS Number:
Molecular Weight:
255.23
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ganciclovir

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC1=Nc2c(ncn2COC(CO)CO)C(=O)N1

InChI

1S/C9H13N5O4/c10-9-12-7-6(8(17)13-9)11-3-14(7)4-18-5(1-15)2-16/h3,5,15-16H,1-2,4H2,(H3,10,12,13,17)

InChI key

IRSCQMHQWWYFCW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ganciclovir EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Muta. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Zhongmin Jiang et al.
International journal of oncology, 57(3), 780-790 (2020-07-25)
It has been controversial whether patients with hepatocellular carcinoma (HCC) should receive glucocorticoid therapy during chemotherapy. Recent studies have demonstrated that glucocorticoids increase the therapeutic sensitivity of tumors to some chemotherapeutic drugs, but the specific mechanism remains unclear. In the
Zheng Z Zhang et al.
Nucleic acids research, 42(21), 13186-13193 (2014-11-08)
The boundaries of R-loops are well-documented at immunoglobulin heavy chain loci in mammalian B cells. Within primary B cells or B cell lines, the upstream boundaries of R-loops typically begin early in the repetitive portion of the switch regions. Most
Chien-Chia Su et al.
American journal of ophthalmology, 158(5), 1024-1031 (2014-08-16)
To evaluate the clinical characteristics and therapeutic outcomes of cytomegalovirus (CMV)-positive Posner-Schlossman syndrome patients undergoing topical ganciclovir treatment. Retrospective, comparative, and interventional case series. One eye of each of 126 consecutive Posner-Schlossman syndrome patients was investigated using aqueous polymerase chain
Fengting Liu et al.
International journal of oncology, 53(5), 2010-2020 (2018-09-19)
Osteosarcoma stem cells are able to escape treatment with conventional chemotherapeutic drugs, as the majority of them are in a quiescent state. Recent reports have suggested that small ubiquitin‑like modifiers (SUMOs) serve important roles in the maintenance of cancer stem cell
Zhuoming Liu et al.
The Journal of experimental medicine, 211(6), 1197-1213 (2014-05-28)
Competition for iron influences host-pathogen interactions. Pathogens secrete small iron-binding moieties, siderophores, to acquire host iron. In response, the host secretes siderophore-binding proteins, such as lipocalin 24p3, which limit siderophore-mediated iron import into bacteria. Mammals produce 2,5-dihydroxy benzoic acid, a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service