Skip to Content
Merck
All Photos(1)

Key Documents

Y0001100

Itraconazole for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Itraconazole, Oriconazole, R51211, Sporanox

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H38Cl2N8O4
CAS Number:
Molecular Weight:
705.63
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

itraconazole

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCC(C)N1N=CN(C1=O)c2ccc(cc2)N3CCN(CC3)c4ccc(OCC5COC(Cn6cncn6)(O5)c7ccc(Cl)cc7Cl)cc4

InChI

1S/C35H38Cl2N8O4/c1-3-25(2)45-34(46)44(24-40-45)29-7-5-27(6-8-29)41-14-16-42(17-15-41)28-9-11-30(12-10-28)47-19-31-20-48-35(49-31,21-43-23-38-22-39-43)32-13-4-26(36)18-33(32)37/h4-13,18,22-25,31H,3,14-17,19-21H2,1-2H3

InChI key

VHVPQPYKVGDNFY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Itraconazole for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Itraconazole inhibits cytochrome P-450-dependent enzymes which results in the inhibition of ergosterol synthesis. It does so by interacting with 14-α demethylase, which is a cytochrome P-450 enzyme necessary to convert lanosterol to ergosterol. Ergosterol is a crucial compenent of fungal cell membranes. Therefore, it′s inhibition results in increased cellular permeability causing leakage of cellular contents. Itraconazole may also inhibit endogenous respiration, interact with membrane phospholipids, inhibit the transformation of yeasts to mycelial forms, inhibit purine uptake, and impair triglyceride and phospholipid biosynthesis.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabriele Sass et al.
The American journal of tropical medicine and hygiene, 101(2), 383-391 (2019-06-21)
Trypanosoma cruzi is the etiologic agent of Chagas disease (CD), which can result in severe cardiomyopathy. Trypanosoma cruzi is endemic to the Americas, and of particular importance in Latin America. In the United States and other non-endemic countries, rising case
C H W Koks et al.
Pharmacological research, 46(2), 195-201 (2002-09-11)
The clinical pharmacology of itraconazole is presented in relation to its use in the treatment of fluconazole-resistant oropharyngeal candidosis. The oral solution is a new formulation of itraconazole in which itraconazole is solubilised with the use of cyclodextrin. This formulation
Anil Kumar et al.
The American journal of tropical medicine and hygiene, 90(4), 735-739 (2014-02-05)
Systemic endemic mycoses, such as blastomycosis, are rare in Asia and have been reported as health risks among travelers who visit or reside in an endemic area. Adrenal involvement is rarely seen in blastomycosis and has never been reported from
Izumi Kikuchi et al.
European journal of dermatology : EJD, 16(1), 42-47 (2006-01-27)
Potassium iodide, itraconazole (ITCZ), and terbinafine are widely known as oral antifungal agents for the treatment of sporotrichosis. Although potassium iodide has been used as the antifungal agent of first choice in Japan due to its high efficacy, its use
James D Homans et al.
The Pediatric infectious disease journal, 29(1), 65-67 (2009-11-04)
Coccidioides immitis causes a wide range of disease in humans. Fluconazole and itraconazole are effective treatments. Clinical evidence suggests that itraconazole is equivalent or superior to fluconazole in treating osteoarticular infections in rates of cure and recurrence. We report 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service