Skip to Content
Merck
All Photos(1)

Key Documents

Y0000853

Fluvoxamine for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Fluvoxamine maleate, (E)-5-Methoxy-1-[4-(trifluoromethyl)phenyl]-1-pentanone-O-(2-aminoethyl)oxime maleate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21F3N2O2 · C4H4O4
CAS Number:
Molecular Weight:
434.41
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

fluvoxamine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COCCCC/C(C1=CC=C(C(F)(F)F)C=C1)=N\OCCN.OC(/C=C\C(O)=O)=O

InChI

1S/C15H21F3N2O2.C4H4O4/c1-21-10-3-2-4-14(20-22-11-9-19)12-5-7-13(8-6-12)15(16,17)18;5-3(6)1-2-4(7)8/h5-8H,2-4,9-11,19H2,1H3;1-2H,(H,5,6)(H,7,8)/b20-14+;2-1-

InChI key

LFMYNZPAVPMEGP-PIDGMYBPSA-N

Gene Information

human ... SLC6A4(6532)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fluvoxamine for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Donatella Marazziti et al.
Human psychopharmacology, 27(4), 397-402 (2012-07-19)
In this study, we explored the possible relationships between plasma fluvoxamine levels and clinical features and/or response in adult obsessive-compulsive disorder (OCD) patients treated with this drug for 6 months. Twenty OCD outpatients of both sexes who were already taking
Shohei Matsunami et al.
Behavioural brain research, 233(2), 577-586 (2012-06-12)
Behavioural effects of fluvoxamine (FLV, selective serotonin reuptake inhibitor) were examined in 1-2 week old domestic chicks. Chicks were tested in an I-shaped maze equipped with a feeder (ON feeder) that served 1 or 2 grains of millet at gradually
Md Shenuarin Bhuiyan et al.
Journal of pharmacological sciences, 121(3), 177-184 (2013-02-23)
Depression is associated with a substantial increase in the risk of developing heart failure and is independently associated with increased cardiovascular morbidity and mortality. Inversely, cardiovascular disease can lead to severe depression. Thus, therapy with selective serotonin reuptake inhibitors (SSRIs)
Natalia Mast et al.
Molecular pharmacology, 82(5), 824-834 (2012-08-04)
Cytochrome P450 46A1 (cholesterol 24-hydroxylase) is an important brain enzyme that may be inhibited by structurally distinct pharmaceutical agents both in vitro and in vivo. To identify additional inhibitors of CYP46A1 among U.S. Food and Drug Administration-approved therapeutic agents, we
Eiko Sunami et al.
Internal medicine (Tokyo, Japan), 51(10), 1187-1193 (2012-06-13)
Antidepressants have been recommended for the treatment of post-stroke depression (PSD). The purpose of this study was to evaluate the effect of fluvoxamine maleate, a selective serotonin re-uptake inhibitor (SSRI), on depressive state, sleep disturbance, and serum melatonin levels in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service