Skip to Content
Merck
All Photos(1)

Key Documents

Y0000662

Ribonucleic acid

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ribonucleic acid EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The RNA revolution.
Christine Gorman et al.
Scientific American, 310(4), 52-59 (2014-04-10)
Xiaoyu Li et al.
Genomics, proteomics & bioinformatics, 12(2), 72-78 (2014-04-22)
RNA-protein interactions influence many biological processes. Identifying the binding sites of RNA-binding proteins (RBPs) remains one of the most fundamental and important challenges to the studies of such interactions. Capturing RNA and RBPs via chemical crosslinking allows stringent purification procedures
Tumour suppressors: Master weaver regulates guardian.
Nicola McCarthy
Nature reviews. Cancer, 14(5), 295-295 (2014-04-18)
The RNA code: nature's Rosetta Stone.
C Thomas Caskey et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(16), 5758-5759 (2014-04-24)
Yann Tonin et al.
The Journal of biological chemistry, 289(19), 13323-13334 (2014-04-03)
Defects in mitochondrial genome can cause a wide range of clinical disorders, mainly neuromuscular diseases. Presently, no efficient therapeutic treatment has been developed against this class of pathologies. Because most of deleterious mitochondrial mutations are heteroplasmic, meaning that wild type

Questions

  1. The pack size of Y0000662 Ribonucleic acid

    1 answer
    1. The package size for this item is 100mg. This product is supplier by the European Pharmacopoeia. For additional information see the link below:
      https://crs.edqm.eu/db/4DCGI/View=Y0000662

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service