Skip to Content
Merck
All Photos(1)

Key Documents

Y0000569

Chlorogenic acid

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1,4,5-Trihydroxycyclohexanecarboxylic acid 3-(3,4-dihydroxycinnamate), 3-(3,4-Dihydroxycinnamoyl)quinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18O9
CAS Number:
Molecular Weight:
354.31
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

chlorogenic acid

manufacturer/tradename

EDQM

mp

210 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O[C@@H]1C[C@](O)(C[C@@H](OC(=O)\C=C\c2ccc(O)c(O)c2)[C@@H]1O)C(O)=O

InChI

1S/C16H18O9/c17-9-3-1-8(5-10(9)18)2-4-13(20)25-12-7-16(24,15(22)23)6-11(19)14(12)21/h1-5,11-12,14,17-19,21,24H,6-7H2,(H,22,23)/b4-2+/t11-,12-,14-,16+/m1/s1

InChI key

CWVRJTMFETXNAD-JUHZACGLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Chlorogenic acid belongs to the class of phenolic acid derivatives commonly found in higher plants.[1] It is widely used as a highly valuable natural polyphenol compound, in consumer chemicals, medicine, food and beverage industries.[2]

Application

Chlorogenic acid EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Characterization of green and roasted coffees through the chlorogenic acid fraction by HPLC-UV and principal component analysis
Bicchi PC, et al.
Journal of Agricultural and Food Chemistry, 43(6), 1549-1555 (1995)
Extraction and HPLC characterization of chlorogenic acid from tobacco residuals
Chen Y, et al.
Separation Science and Technology, 42(15), 3481-3492 (2007)
Jing Chen et al.
Journal of chromatography. A, 1352, 55-61 (2014-06-11)
The overlapping and shifts of peaks and noise signals appear mostly in high performance liquid chromatography (HPLC) experiments. A practical application of wavelet moment method on the quantitative analysis of the main active components in Shuanghuanglian oral liquid samples was
Marta González-Castejón et al.
Phytotherapy research : PTR, 28(5), 745-752 (2013-08-21)
In this in vitro study, we have investigated the ability of Taraxacum officinale (dandelion) to inhibit adipocyte differentiation and lipogenesis in 3T3-L1 preadipocytes. HPLC analysis of the three plant extracts used in this study-leaf and root extracts and a commercial
Senem Kamiloglu et al.
Journal of the science of food and agriculture, 94(11), 2225-2233 (2014-01-01)
In order to investigate the effect of home processing on the bioaccessibility of health-related constituents of tomatoes, total lycopene, phenolics, flavonoids and antioxidant capacity were determined from seven different tomato products using an in vitro gastrointestinal digestion model. Additionally, the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service