Skip to Content
Merck
All Photos(1)

Key Documents

Y0000500

Acarbose

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Acarbose, 4",6"-Dideoxy-4"-([1S]-[1,4,6/5]-4,5,6-trihydroxy-3-hydroxymethyl-2-yclohexenylamino)-maltotriose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H43NO18
CAS Number:
Molecular Weight:
645.60
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

Agency

EP Reference Standard

API family

acarbose

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O[C@@H]2CO)O[C@H]3[C@H](O)[C@@H](O)[C@H](O)O[C@@H]3CO)[C@H](O)[C@@H](O)[C@@H]1N[C@H]4C=C(CO)[C@@H](O)[C@H](O)[C@H]4O

InChI

1S/C25H43NO18/c1-6-11(26-8-2-7(3-27)12(30)15(33)13(8)31)14(32)19(37)24(40-6)43-22-10(5-29)42-25(20(38)17(22)35)44-21-9(4-28)41-23(39)18(36)16(21)34/h2,6,8-39H,3-5H2,1H3/t6-,8+,9-,10-,11-,12-,13+,14+,15+,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-/m1/s1

InChI key

XUFXOAAUWZOOIT-SXARVLRPSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Acarbose EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Modified tetrasaccharide that acts as a reversible α -glucosidase inhibitor.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun-Peng Yang et al.
Journal of agricultural and food chemistry, 62(31), 7760-7770 (2014-07-16)
Bamboo leaf extract as a food additive has been used for preventing the oxidation of food. In the present study, we investigated the influence of Phyllostachys edulis leaf extract on starch digestion. Orientin, isoorientin, vitexin, and isovitexin were determined as
Sophie Delorme et al.
Current opinion in pharmacology, 5(2), 184-189 (2005-03-23)
The prevalence of glucose intolerance is increasing dramatically worldwide. Both impaired glucose tolerance (IGT) and type 2 diabetes are associated with excess mortality from cardiovascular disease. It is now generally accepted that these cardiovascular complications are related to prevailing hyperglycemia
Matteo Monami et al.
Advances in therapy, 29(9), 736-746 (2012-08-28)
Lipid profile is an important determinant of cardiovascular risk in type 2 diabetes. It is well known that patients with type 2 diabetes are more likely to be dyslipidemic than the general population. Given the observed connection between glucose and
C Deliens et al.
Acta clinica Belgica, 69(4), 290-293 (2014-06-12)
Postprandial reactive hypoglycaemia (PRH) is a clinical syndrome characterized by the recurrence of symptomatic hypoglycaemia during postprandial periods. PRH remains a diagnostic challenge for clinicians, because of its atypical manifestations and low prevalence, especially in older persons. We report the
Uwe Zeymer
International journal of cardiology, 107(1), 11-20 (2005-12-13)
Impaired glucose metabolism is associated with an increased risk of cardiovascular events and cardiovascular-associated mortality. Postprandial hyperglycaemia is one of the earliest identifiable indicators of impaired glucose control. It contributes to the progression from impaired glucose tolerance to overt type

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service