Skip to Content
Merck
All Photos(1)

Key Documents

Y0000429

Trospium chloride

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H30ClNO3
CAS Number:
Molecular Weight:
427.96
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

trospium

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Cl-].[N+]21(C3CCC2CC(C3)OC(=O)C(O)(c5ccccc5)c4ccccc4)CCCC1

InChI

1S/C25H30NO3.ClH/c27-24(25(28,19-9-3-1-4-10-19)20-11-5-2-6-12-20)29-23-17-21-13-14-22(18-23)26(21)15-7-8-16-26;/h1-6,9-12,21-23,28H,7-8,13-18H2;1H/q+1;/p-1

InChI key

RVCSYOQWLPPAOA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Trospium chloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anthony G Visco et al.
The New England journal of medicine, 367(19), 1803-1813 (2012-10-06)
Anticholinergic medications and onabotulinumtoxinA are used to treat urgency urinary incontinence, but data directly comparing the two types of therapy are needed. We performed a double-blind, double-placebo-controlled, randomized trial involving women with idiopathic urgency urinary incontinence who had five or
Mark D Harnett et al.
Clinical drug investigation, 33(2), 133-141 (2012-12-04)
Overactive bladder is a common disorder that affects approximately 34 million adults in the United States. Anticholinergic (antimuscarinic) agents are the most widely used pharmacological option for overactive bladder. This study set out to identify and characterize the influence of
Norman R Zinner et al.
Neurourology and urodynamics, 30(7), 1214-1219 (2011-04-05)
Once-daily extended-release (XR) trospium chloride has been evaluated for the treatment of overactive bladder syndrome (OAB) in two 12-week randomized, double-blind, placebo-controlled studies. This pooled analysis of the 9-month open-label extensions to these studies evaluated the long-term efficacy and tolerability
David A Ginsberg et al.
Neurourology and urodynamics, 30(4), 563-567 (2011-01-27)
Once-daily extended release (XR) trospium chloride, which provides therapeutic trospium plasma concentrations over 24 hours, has demonstrated efficacy in treating overactive bladder (OAB) symptoms as evaluated over a 24-hr period. This analysis examined the effects of trospium XR on diurnal
Peter K Sand et al.
Drugs & aging, 28(2), 151-160 (2011-02-01)
Overactive bladder syndrome (OAB) is associated with various co-morbidities; treatment of these frequently results in multiple medication use (MMU) and the potential for drug-drug interactions, which may lead to adverse events and altered efficacy. With the aging population, the prevalence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service