Skip to Content
Merck
All Photos(1)

Documents

Y0000196

Ondansetron for TLC system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Ondansetron hydrochloride dihydrate, 1,2,3,9-Tetrahydro-9-methyl-3-[(2-methyl-1H-imidazol-1-yl)methyl]-4H-carbazol-4-one hydrochloride, GR 38032F

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19N3O · HCl · 2H2O
CAS Number:
Molecular Weight:
365.85
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ondansetron

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].[H]O[H].[H]O[H].Cc1nccn1CC2CCc3c(C2=O)c4ccccc4n3C

InChI

1S/C18H19N3O.ClH.2H2O/c1-12-19-9-10-21(12)11-13-7-8-16-17(18(13)22)14-5-3-4-6-15(14)20(16)2;;;/h3-6,9-10,13H,7-8,11H2,1-2H3;1H;2*1H2

InChI key

VRSLTNZJOUZKLX-UHFFFAOYSA-N

Gene Information

human ... HTR3A(3359)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ondansetron for TLC system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

5-HT3 serotonin receptor antagonist

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristi L Kinder et al.
Pediatric emergency care, 28(11), 1203-1205 (2012-11-02)
The objective of this study was to determine if overweight children are more likely than normal-weight children to require ondansetron when undergoing ketamine sedation in a pediatric emergency department. Patients between the ages of 2 and 18 years with an
Mehrdad Kashifard et al.
Clinical and experimental obstetrics & gynecology, 40(1), 127-130 (2013-06-04)
Nausea and vomiting of pregnancy (NVP) are seen in 50-80% of pregnancies. However, in severe NVP, called hypermesis gravidarum (HG), medical therapy to reduce nausea and vomiting is inevitable and ondansetron (OND) as an effective drug has recently been proposed.
Daniel Hervás et al.
Pediatric emergency care, 28(11), 1166-1168 (2012-11-02)
In this study, we determine the clinical impact of 1 dose of oral ondansetron for children with vomiting and evaluate the economic consequences of its use. The strategies compared were administering oral ondansetron in addition to oral rehydration therapy (group
Hitendra S Mahajan et al.
Carbohydrate polymers, 91(2), 618-625 (2012-11-06)
The objective of present study was to enhance bioadhesive potential of xyloglucan by thiolation. Thiolation of xyloglucan was achieved with esterification with thioglycolic acid. Thiolated xyloglucan was characterized by NMR, DSC, and XRD analysis. Thiolated xyloglucan was determined to possess
João Maria Corrêa Filho et al.
Addictive behaviors, 38(4), 2044-2051 (2013-02-12)
Ondansetron has emerged as a promising medication for the treatment of alcohol dependence, mainly among early-onset alcoholics. This research primarily aimed to evaluate the efficacy and safety of ondansetron at a 16mg/day dosage to treat alcohol-dependent outpatients. A double-blind, placebo-controlled

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service