Skip to Content
Merck
All Photos(1)

Key Documents

Y0000193

Carteolol for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Carteolol monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24N2O3 · HCl
CAS Number:
Molecular Weight:
328.83
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

carteolol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Cl-].N1c2c(c(ccc2)OCC(O)CNC(C)(C)C)CCC1=O.[H+]

InChI

1S/C16H24N2O3.ClH/c1-16(2,3)17-9-11(19)10-21-14-6-4-5-13-12(14)7-8-15(20)18-13;/h4-6,11,17,19H,7-10H2,1-3H3,(H,18,20);1H

InChI key

FYBXRCFPOTXTJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Carteolol for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maura Floreani et al.
European journal of pharmacology, 590(1-3), 269-275 (2008-06-24)
This in vitro study was designed to investigate whether carteolol, a non-conventional partial agonists of beta(1)-adrenoceptors, relaxes phenylephrine-constricted rat aorta through activation of the low-affinity state of beta(1)-adrenoceptors or antagonist effect at alpha(1)-adrenoceptors. Carteolol-induced complete concentration-dependent relaxation of phenylephrine-contracted aorta
Tetsuya Yamamoto
Nippon Ganka Gakkai zasshi, 111(6), 463-472 (2007-07-03)
To compare the ocular hypotensive activity and safety profile of long-acting 1% carteolol hydrochloride eye drops (long-acting formulation) to those of 1% carteolol hydrochloride eye drops(currently prescribed drug) for reduction of intraocular pressure. Patients with primary open-angle glaucoma or ocular
The possible treatment of migraine with carteolol hydrochloride.
M K Shapiro et al.
Headache, 39(2), 138-139 (2008-06-21)
Tetsuya Sugiyama et al.
Acta ophthalmologica, 87(7), 797-800 (2008-12-05)
To assess the effects of combined therapy with latanoprost and beta blockers on optic nerve head (ONH) blood flow in normal-tension glaucoma (NTG) patients. Intraocular pressure (IOP), ONH blood flow (laser speckle flowgraphy) and blood pressure were measured in 15
Sheridan Henness et al.
Drugs & aging, 24(6), 509-528 (2007-06-19)
Ocular carteolol (Mikelan), Teoptic, Ocupress) is a nonselective beta-adrenoceptor antagonist with intrinsic sympathomimetic activity (ISA). Ocular carteolol effectively reduces intraocular pressure (IOP) in patients with open-angle glaucoma (OAG) or ocular hypertension (OH). Twice-daily administration of standard carteolol has generally similar

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service