Skip to Content
Merck
All Photos(1)

Key Documents

Y0000079

Articaine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-Methyl-3-[[1-oxo-2-(propylamino)propyl]amino]-2-thiophenecarboxylic acid methyl ester hydrochloride, Carticaine chlorhydrate, Carticaine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20N2O3S · HCl
CAS Number:
Molecular Weight:
320.84
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

articaine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC1=CSC(C(OC)=O)=C1NC(C(C)NCCC)=O.Cl

InChI

1S/C13H20N2O3S.ClH/c1-5-6-14-9(3)12(16)15-10-8(2)7-19-11(10)13(17)18-4;/h7,9,14H,5-6H2,1-4H3,(H,15,16);1H

InChI key

GDWDBGSWGNEMGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Articaine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Articaine is an amide local anesthetic widely used in dentistry. Articaine acts by inhibition of nerve impulse conduction by blockade of sodium channels.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kagan Degerliyurt et al.
Implant dentistry, 22(3), 289-294 (2013-05-08)
Although epinephrine is one of the most commonly used vasoconstrictor in association with local anesthesia in dentistry, systemic effects of topical admission of epinephrine for sinus augmentation have not been investigated yet. The purpose of this study was to reveal
Urticaria due to an intradermal test with articaine hydrochloride.
G Davila-Fernández et al.
Journal of investigational allergology & clinical immunology, 22(5), 373-374 (2012-10-30)
R Leith et al.
European archives of paediatric dentistry : official journal of the European Academy of Paediatric Dentistry, 13(6), 293-296 (2012-12-14)
Lidocaine has been considered the gold standard for local analgesia agents in dentistry for years. Articaine is now widely used but there has been a reluctance to use it in children. Compared with lidocaine, articaine is 1.5 times as potent
Sepideh Amjad-Iranagh et al.
Journal of molecular modeling, 19(9), 3831-3842 (2013-06-27)
Articaine, as a local anesthetic drug has been simulated in neutral and charged forms, and its interaction with the dimyristoylphosphatidylcholine (DMPC) lipid bilayer membrane is investigated by molecular dynamics simulation using GROMACS software. In order to obtain the optimum location
Hécio Henrique Araújo de Morais et al.
The Journal of craniofacial surgery, 23(6), 1703-1708 (2012-11-14)
The aim of the present study was to analyze hemodynamic changes after the administration of either 2% lidocaine with epinephrine 1:100,000 (L100) or 4% articaine with epinephrine 1:200,000 (A200) in the surgical removal of symmetrically positioned lower third molars. A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service