Skip to Content
Merck
All Photos(1)

Key Documents

Y0000073

Cetyl palmitate 15

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Palmityl palmitate, Cetyl palmitate, Hexadecyl hexadecanoate, Palmitic acid palmityl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14CO2(CH2)15CH3
CAS Number:
Molecular Weight:
480.85
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

cetyl palmitate

manufacturer/tradename

EDQM

mp

55-56 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCC

InChI

1S/C32H64O2/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-34-32(33)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-31H2,1-2H3

InChI key

PXDJXZJSCPSGGI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cetyl palmitate 15 EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparative study of organic solvent and water-soluble lipophilic extractives from wheat straw I: yield and chemical composition.
Sun C R, et al.
Journal of wood science, 49(1), 0047-0052 (2003)
Fundamentals of Environmental and Toxicological Chemistry: Sustainable Science, Fourth Edition (2013)
Fundamentals of Environmental and Toxicological Chemistry: Sustainable Science, Fourth Edition (2013)
Ehsan Aboutaleb et al.
Journal of biomedical materials research. Part A, 102(7), 2125-2136 (2013-07-31)
Vincristine (VC) sulfate, a freely water-soluble cytotoxic agent was incorporated into cetyl palmitate solid lipid nanoparticles (SLNs) with the aid of dextran sodium sulfate (DS), a poly anion, using a microemulsion method. The manufacturing process was optimized using response surface
Silke B Lohan et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 89, 201-207 (2014-12-17)
UV irradiation leads to the formation of reactive oxygen species (ROS). An imbalance between the antioxidant system and ROS can lead to cell damage, premature skin aging or skin cancer. To counteract these processes, antioxidants such as coenzyme Q10 (CoQ10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service