All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H12O2
CAS Number:
Molecular Weight:
152.19
FEMA Number:
4192
MDL number:
UNSPSC Code:
12164502
Flavis number:
13.163
NACRES:
NA.21
Organoleptic:
licorice; cheesy; earthy; sweet
grade:
FG
Agency:
meets purity specifications of JECFA
food allergen:
no known allergens
Recommended Products
grade
FG
Quality Level
Agency
meets purity specifications of JECFA
reg. compliance
EU 1334/2008 & 178/2002
FDA 21 CFR 117
Assay
≥99%
application(s)
flavors and fragrances
food allergen
no known allergens
Organoleptic
licorice; cheesy; earthy; sweet
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Skin Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service