Skip to Content
Merck
All Photos(7)

Key Documents

T8625

Sigma-Aldrich

L-Threonine

≥98% (HPLC)

Synonym(s):

(2S,3R)-2-Amino-3-hydroxybutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH(NH2)CO2H
CAS Number:
Molecular Weight:
119.12
Beilstein:
1721646
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Product Name

L-Threonine, reagent grade, ≥98% (HPLC)

grade

reagent grade

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

mp

256 °C (dec.) (lit.)

solubility

H2O: 50 mg/mL, clear, colorless

application(s)

cell analysis

SMILES string

C[C@@H](O)[C@H](N)C(O)=O

InChI

1S/C4H9NO3/c1-2(6)3(5)4(7)8/h2-3,6H,5H2,1H3,(H,7,8)/t2-,3+/m1/s1

InChI key

AYFVYJQAPQTCCC-GBXIJSLDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-threonine has been used for the selection of transformed cells. It has also been used to study the effects of amino acids in promoting food consumption in Drosophila melanogaster.

Biochem/physiol Actions

L-Threonine is an essential amino acid. It is involved in the synthesis of mucin, a protein supporting intestinal function and integrity. L-threonine has a role in the immune system, in O-linked glycosylation, protein phosphorylation and glycine synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Markerless Gene Editing in the Hyperthermophilic Archaeon Thermococcus kodakarensis
Alexandra M. Gehring
Bio-protocol, 7(22), 1-26 (2017)
An internal sensor detects dietary amino acids and promotes food consumption in Drosophila
Zhe Yang
Nature, 1-36 (2017)
Yongxian Zhuang et al.
PloS one, 9(9), e108444-e108444 (2014-09-26)
Different cancer cells exhibit altered sensitivity to metformin treatment. Recent studies suggest these findings may be due in part to the common cell culture practice of utilizing high glucose, and when glucose is lowered, metformin becomes increasingly cytotoxic to cancer
Tamara Seredenina et al.
Antioxidants & redox signaling, 23(5), 490-513 (2014-02-04)
Voltage-gated proton channels are increasingly implicated in cellular proton homeostasis. Proton currents were originally identified in snail neurons less than 40 years ago, and subsequently shown to play an important auxiliary role in the functioning of reactive oxygen species (ROS)-generating
Mario Ost et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(4), 1314-1328 (2014-12-11)
Recent studies on mouse and human skeletal muscle (SM) demonstrated the important link between mitochondrial function and the cellular metabolic adaptation. To identify key compensatory molecular mechanisms in response to chronic mitochondrial distress, we analyzed mice with ectopic SM respiratory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service