Skip to Content
Merck
All Photos(1)

Key Documents

T7511

Supelco

Tripelennamine hydrochloride

analytical standard

Synonym(s):

2-(Benzyl[2-dimethylaminoethyl]amino)pyridine, N-Benzyl-N′,N′-dimethyl-N-(2-pyridyl)ethylenediamine hydrochloride, Pyribenzamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H21N3 · HCl
CAS Number:
Molecular Weight:
291.82
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
forensics and toxicology
personal care
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

CN(C)CCN(CC1=CC=CC=C1)C2=NC=CC=C2.Cl

InChI

1S/C16H21N3.ClH/c1-18(2)12-13-19(16-10-6-7-11-17-16)14-15-8-4-3-5-9-15;/h3-11H,12-14H2,1-2H3;1H

InChI key

FSSICIQKZGUEAE-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

General description

Tripelennamine hydrochloride is an antihistamine drug, which is used in the treatment of bronchitis, asthma and bronchoconstriction.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Tripelennamine hydrochloride may be used as a reference standard in the determination of tripelennamine hydrochloride in samples of animal feed by gas chromatography (GC) method, wastewater and urine using high performance liquid chromatography (HPLC) method employing fluorescence detector.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trace determination of the antihistamines tripelennamine hydrochloride, thenyldiamine hydrochloride, and chlorothen citrate in admixture in animal feed, human urine, and wastewater by high-pressure liquid chromatography and use of a fluorescence detector.
Thompson TH, et al.
Talanta, 30(4), 251-260 (1983)
Ann E Stuart et al.
Visual neuroscience, 19(3), 307-319 (2002-10-24)
Using autoradiographic and biochemical techniques, we studied the sodium-dependent forward and reverse transport of the neurotransmitter histamine in an arthropod photoreceptor in order to test whether the transporter plays a central role in visual signal transfer at this synapse. In
Shilpa P Sakhalkar et al.
International immunopharmacology, 5(7-8), 1299-1309 (2005-05-26)
Signal transducer and activator of transcription-1 (STAT1) is a latent signal transducer protein which, on phosphorylation, is translocated from the cytoplasm to the nucleus and is subsequently activated. This study was designed to determine the involvement of histamine receptors in
T Sato et al.
Japanese journal of pharmacology, 71(1), 81-84 (1996-05-01)
Effects of histamine H1-receptor antagonists on L-dopa-induced behavioral excitement were examined in mice to confirm behaviorally the inhibition of dopamine uptake by these compounds. L-Dopa (100-300 mg/kg, s.c.) combined with pargyline hydrochloride (80 mg/kg, i.p.) caused a dose-dependent behavioral excitement.
Ulrich Schlecht et al.
PloS one, 7(8), e42853-e42853 (2012-08-21)
Meiosis is a highly regulated developmental process that occurs in all eukaryotes that engage in sexual reproduction. Previous epidemiological work shows that male and female infertility is rising and environmental factors, including pollutants such as organic solvents, are thought to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service