Skip to Content
Merck
All Photos(4)

Key Documents

T35955

Sigma-Aldrich

p-Toluenesulfonyl chloride

reagent grade, ≥98%

Synonym(s):

Tosyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO2Cl
CAS Number:
Molecular Weight:
190.65
Beilstein:
607898
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

reagent grade

Quality Level

vapor pressure

1 mmHg ( 88 °C)

Assay

≥98%

bp

134 °C/10 mmHg (lit.)

mp

65-69 °C (lit.)

SMILES string

Cc1ccc(cc1)S(Cl)(=O)=O

InChI

1S/C7H7ClO2S/c1-6-2-4-7(5-3-6)11(8,9)10/h2-5H,1H3

InChI key

YYROPELSRYBVMQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

p-toluenesulfonyl chloride may be used along with N-methylimidazole for the esterification or thioesterification of carboxylic acids and alcohols or thiols. It may also be used as a chlorine source for α-chlorination of ketones in the presence of lithium diisopropylamide.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

262.4 °F - closed cup

Flash Point(C)

128 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rince Wong et al.
The Journal of organic chemistry, 72(10), 3969-3971 (2007-04-21)
A facile and general protocol for the preparation of isothiocyanates from alkyl and aryl amines is reported. This method relies on a tosyl chloride mediated decomposition of a dithiocarbamate salt that is generated in situ by treatment of an amine
Katarzyna Dziarkowska et al.
Analytica chimica acta, 606(2), 184-193 (2007-12-18)
Determination of polyamines in biological fluids possesses medical diagnostic relevance. Despite the vast panel of analytical methods developed for polyamines they are not applied in routine clinical usage, mainly due to the time and labor consuming sample preparation step and
Salvatore Lombardo et al.
Langmuir : the ACS journal of surfaces and colloids, 33(22), 5473-5481 (2017-05-13)
The interaction of bovine serum albumin (BSA) with sulfated, carboxylated, and pyridinium-grafted cellulose nanocrystals (CNCs) was studied as a function of the degree of substitution by determining the adsorption isotherm and by directly measuring the thermodynamics of interaction. The adsorption
Y Tabata et al.
Biomaterials, 7(3), 234-238 (1986-05-01)
The cyanogen bromide (CNBr) activation method was adopted to link collagen molecules onto the surface of cellulose and poly(vinyl alcohol) (PVA) films via covalent bonding. The amount of bound protein was determined by the ninhydrin method and found to be
I M Kung et al.
Biomaterials, 16(8), 649-655 (1995-05-01)
Three different surface modifications were conducted on the membranes of double-layer alginate/poly(L-lysine) microcapsules with tosyl chloride-activated poly(ethylene glycol), cyanuric chloride-activated poly(ethylene glycol) and tosyl chloride-activated poly(vinyl alcohol), separately. All these surface modifications strengthen the microcapsular membranes. Of the three surface-modified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service