Skip to Content
Merck
All Photos(1)

Key Documents

T1899990

Triamcinolone

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

9α-Fluoro-11β,16α,17,21-tetrahydroxy-1,4-pregnadiene-3,20-dione, 9α-Fluoro-11β,16α,17,21-tetrahydroxypregna-1,4-diene-3,20-dione, Fluoxyprednisolone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27FO6
CAS Number:
Molecular Weight:
394.43
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

triamcinolone

manufacturer/tradename

EDQM

mp

262-263 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12C[C@@H](O)[C@](O)(C(=O)CO)[C@@]1(C)C[C@H](O)[C@@]3(F)[C@@]2([H])CCC4=CC(=O)C=C[C@]34C

InChI

1S/C21H27FO6/c1-18-6-5-12(24)7-11(18)3-4-13-14-8-15(25)21(28,17(27)10-23)19(14,2)9-16(26)20(13,18)22/h5-7,13-16,23,25-26,28H,3-4,8-10H2,1-2H3/t13-,14-,15+,16-,18-,19-,20-,21-/m0/s1

InChI key

GFNANZIMVAIWHM-OBYCQNJPSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Triamcinolone EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Triamcinolone is a synthetic glucocorticoid agonist; induces gene expression and apoptosis; inhibits prostaglandin synthesis; impairs tumor necrosis factor (TNF)-α-induced degradation of κB-α; potentiates the differentiation-inducing effects of bone morphogenetic proteins (BMP-2, -4, -6).

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hypopigmentation along subcutaneous veins following intrakeloid triamcinolone injection: a case report and review of literature.
P Kumar et al.
Burns : journal of the International Society for Burn Injuries, 24(5), 487-488 (1998-09-02)
J C Santoso-Pham et al.
Cutis, 59(6), 329-332 (1997-06-01)
Keratoacanthoma is a common skin tumor characterized by rapid growth of a smooth dome-shaped nodule with a central plug of keratin, usually followed by spontaneous involution. We report the case of a giant keratoacanthoma on the face that continued to
Jacob Karsh et al.
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology, 90(2), 254-258 (2003-02-27)
The primary objective was to report a case of triamcinolone-induced anaphylaxis and review the proposed mechanisms of corticosteroid-associated hypersensitivity reactions. Articles in French and English were identified from references in relevant articles and from articles retrieved from the PubMed web
Jorie A Glick Frasiolas et al.
The Annals of pharmacotherapy, 44(10), 1641-1646 (2010-08-26)
To review published literature regarding the safety and effectiveness of intrapericardial triamcinolone for the treatment of autoreactive pericarditis. Searches of MEDLINE (1966-June 2010) and Cochrane Database (1993-June 2010) were conducted. Limits included articles published in English reporting on human subjects.
Pseudohypopyon after intravitreal triamcinolone injection for the treatment of pseudophakic cystoid macular oedema.
S D M Chen et al.
The British journal of ophthalmology, 88(6), 843-844 (2004-05-19)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service