Skip to Content
Merck
All Photos(1)

Key Documents

T0071000

Terfenadine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

α-[4-(1,1-Dimethylethyl)phenyl]-4-(hydroxydiphenylmethyl)-1-piperidinebutanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H41NO2
CAS Number:
Molecular Weight:
471.67
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

terfenadine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C)(C)c1ccc(cc1)C(O)CCCN2CCC(CC2)C(O)(c3ccccc3)c4ccccc4

InChI

1S/C32H41NO2/c1-31(2,3)26-18-16-25(17-19-26)30(34)15-10-22-33-23-20-29(21-24-33)32(35,27-11-6-4-7-12-27)28-13-8-5-9-14-28/h4-9,11-14,16-19,29-30,34-35H,10,15,20-24H2,1-3H3

InChI key

GUGOEEXESWIERI-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Terfenadine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Non-sedating second generation H1 histamine receptor antagonist.
Non-sedating second generation H1 histamine receptor antagonist. Mainly metabolized by Cyp3A4, 5, 7. Inhibits CYP2C8.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Boram Lee et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 70, 94-99 (2014-05-03)
CYP2J2 enzyme is highly expressed in human tumors and carcinoma cell lines, and epoxyeicosatrienoic acids, CYP2J2-mediated metabolites, have been implicated in the pathologic development of human cancers. To identify a CYP2J2 inhibitor, 50 natural products obtained from plants were screened
Hikaru Tanaka et al.
Biological & pharmaceutical bulletin, 37(11), 1826-1830 (2014-11-05)
The effects of terfenadine and pentamidine on the human ether-a-go-go related gene (hERG) channel current and its intracellular trafficking were evaluated. Green fluorescent protein (GFP)-linked hERG channels were expressed in HEK293 cells, and the membrane current was measured by an
K Kasutani et al.
British journal of pharmacology, 171(22), 5049-5058 (2014-06-20)
IL-31, which is described as a pruritogenic cytokine, is linked to the itching that is associated with allergic and non-allergic eczema, but the precise pruritogenic mechanism of IL-31 and its potential as a therapeutic target for atopic dermatitis (AD) have
Katja Pajula et al.
Molecular pharmaceutics, 11(7), 2271-2279 (2014-05-16)
Combinatorial chemistry has enabled the production of very potent drugs that might otherwise suffer from poor solubility and low oral bioavailability. One approach to increase solubility is to make the drug amorphous, which leads to problems associated with drug stability.
Minjoo Kim et al.
PloS one, 10(3), e0119519-e0119519 (2015-03-18)
Existing data on the association between being overweight and cardiovascular morbidity and mortality risk in adults are inconsistent. We prospectively and longitudinally investigated the effects of weight on arterial stiffness and plasma metabolites in middle-aged subjects (aged 40-55 years). A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service