Skip to Content
Merck
All Photos(1)

Key Documents

T0060000

Terconazole

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(+-)-1-{4-[cis-2-(2,4-Dichlorophenyl)-2-(1H-1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-ylmethoxy]phenyl}-4-isopropylpiperazine, Triaconazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H31Cl2N5O3
CAS Number:
Molecular Weight:
532.46
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

terconazole

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Clc1c(ccc(c1)Cl)[C@@]3(O[C@H](CO3)COc4ccc(cc4)N5CCN(CC5)C(C)C)C[n]2ncnc2

InChI

1S/C26H31Cl2N5O3/c1-19(2)31-9-11-32(12-10-31)21-4-6-22(7-5-21)34-14-23-15-35-26(36-23,16-33-18-29-17-30-33)24-8-3-20(27)13-25(24)28/h3-8,13,17-19,23H,9-12,14-16H2,1-2H3/t23-,26-/m0/s1

InChI key

BLSQLHNBWJLIBQ-OZXSUGGESA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Terconazole EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R G García Figueroa et al.
Ginecologia y obstetricia de Mexico, 68, 154-159 (2000-05-29)
A multicenter randomized study was performed. One hundred and seventy patients were selected. The patients were 18 years and older. They presented signs and symptoms of genital candidiasis and had positive smear culture for Candida. Eighty five patients were assigned
C Schmitt et al.
Obstetrics and gynecology, 76(3 Pt 1), 414-416 (1990-09-01)
Terconazole is the first topical triazole antifungal agent. The results of several European clinical trials have shown both terconazole cream and suppositories to be effective and safe in the treatment of vulvovaginal candidiasis. We conducted a comparative placebo-controlled, double-blind study
C R Cooper et al.
American journal of obstetrics and gynecology, 175(6), 1626-1631 (1996-12-01)
Fifty clinical yeast isolates, representing equally Candida albicans, Candida krusei, Candida parapsilosis, Candida tropicalis, and Torulopsis glabrata, were tested in vitro for their susceptibility to terconazole and fluconazole. The minimal inhibitory concentrations of terconazole and fluconazole were determined by use
J Van Cutsem
American journal of obstetrics and gynecology, 165(4 Pt 2), 1200-1206 (1991-10-01)
The activity of the broad-spectrum triazole antifungal terconazole was evaluated in vitro by the serial decimal dilution technique in broth media. The best correlation between in vitro and in vivo activity was found in brain-heart infusion broth and Eagle's minimum
Fazal Shirazi et al.
Antimicrobial agents and chemotherapy, 57(9), 4360-4368 (2013-07-03)
The high mortality rate of mucormycosis with currently available monotherapy has created interest in studying novel strategies for antifungal agents. With the exception of amphotericin B (AMB), the triazoles (posaconazole [PCZ] and itraconazole [ICZ]) are fungistatic in vitro against Rhizopus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service