Skip to Content
Merck
All Photos(3)

Key Documents

STS0214

Sigma-Aldrich

ECO BRIJ® O20

greener alternative

average Mn ~1,150

Synonym(s):

BRIJ® O20, Polyoxyethylene (20) oleyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C18H35(OCH2CH2)nOH, n~20
CAS Number:
Molecular Weight:
1149.53
MDL number:
UNSPSC Code:
12165104
NACRES:
NA.28

description

non-ionic

Quality Level

form

solid

mol wt

average Mn ~1,150

greener alternative product characteristics

Use of Renewable Feedstocks
Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

25-30 °C (lit.)

hydroxyl value

50‑65 mg KOH/g

solubility

water: soluble 1 g/L at 20 °C

density

0.901 g/cm3 at 20 °C

HLB

15.5

greener alternative category

SMILES string

O(CCO)CCCCCCCC\C=C/CCCCCCCC

InChI

1S/C20H40O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19-22-20-18-21/h9-10,21H,2-8,11-20H2,1H3/b10-9-

InChI key

KWVPFECTOKLOBL-KTKRTIGZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

ECO Brij® O20, also known as oleyl alcohol polyoxyethylene ether, is a bio-based, high HLB nonionic surfactant manufactured from naturally occurring straight-chain oleyl alcohol. This detergent, derived from natural sources, provides various functional advantages, such as detergency, emulsification, and wetting, making it suitable for a range of applications in biochemical and biological research.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is a biobased surfactant and is aligned with the 7th principle of Green Chemistry "Use of Renewable Feedstocks" and the 10th principle "Design for Degradation".

Features and Benefits

  • 100 % Renewable
  • 100 % Bio-based
  • Certified to the USDA BioPreferred Program
  • Lower carbon footprint than petrochemical-based versions
  • High-purity chemical suitable for a wide variety of research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

Brij is a registered trademark of Croda International PLC
ECO BRIJ is a registered trademark of Croda Inc.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

>464.0 °F - Equilibrium method

Flash Point(C)

> 240 °C - Equilibrium method


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yash Kapoor et al.
International journal of pharmaceutics, 361(1-2), 222-229 (2008-06-26)
Cyclosporine A (CyA) is an immunosuppressant drug that is used for treating a variety of ocular diseases and disorders. CyA is commonly delivered via eye drops, which is highly inefficient due to a low bioavailability of less than 5%. The
R B Shah et al.
International journal of pharmaceutics, 341(1-2), 189-194 (2007-05-25)
In the present work, a novel application of ultrasonic measurements is detailed to characterize nano-emulsion formulations as a part of the overall Quality by Design (QbD) goal. Ultrasonic resonator technology (URT) was utilized to measure sound velocity and absorption of
Myriam Chentouf et al.
Journal of immunology (Baltimore, Md. : 1950), 179(1), 409-420 (2007-06-21)
The biological effects of rIgG(1) 13B8.2, directed against the CDR3-like loop on the D1 domain of CD4, are partly due to signals that prevent NF-kappaB nuclear translocation, but the precise mechanisms of action, particularly at the level of membrane proximal
Carles Gil et al.
Biochemical and biophysical research communications, 348(4), 1334-1342 (2006-08-22)
Although the high presence of cholesterol in nerve terminals is well documented, specific roles of this lipid in transmitter release have remained elusive. Since cholesterol is a highly enriched component in the membrane microdomains known as lipid rafts, it is
Sergey Filippov et al.
Langmuir : the ACS journal of surfaces and colloids, 24(17), 9295-9301 (2008-08-09)
In this work we report a new type of pH-responsive micelle-like nanoparticle. Reversible nanoscale structures are formed in solutions of a pH-sensitive hydrophobic polyelectrolyte, poly( N-methacryloyl- l-valine) or poly( N-methacryloyl- l-phenylalanine), and nonionic surfactant (Brij 98) in the presence of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service