S2388
Starch from corn
for use with Total Dietary Fiber Control Kit, TDF-C10
Synonym(s):
Corn starch, Starch from maize
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(C6H10O5)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:
NACRES:
NA.21
Recommended Products
quality
for use with Total Dietary Fiber Control Kit, TDF-C10
Quality Level
technique(s)
photometry: suitable
application(s)
food and beverages
general analytical
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
Internal control for use with the Total Dietary Fiber Assay Kit.
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Reply to J.E. Galgani and G. Valentino.
Coby Eelderink et al.
The American journal of clinical nutrition, 97(4), 903-904 (2013-05-01)
Should insulin resistance degree be taken into account for assessment of glycemic index?
Jose E Galgani et al.
The American journal of clinical nutrition, 97(4), 902-903 (2013-03-22)
Evolutionary genomics: Detecting selection.
Gregory S Barsh et al.
Nature, 495(7441), 325-326 (2013-03-23)
Ayse Bengisu Seferoglu et al.
Plant science : an international journal of experimental plant biology, 205-206, 29-37 (2013-03-19)
ADP-glucose pyrophosphorylase (AGPase) is a key enzyme in plant starch biosynthesis. It contains large (LS) and small (SS) subunits encoded by two different genes. In this study, we explored the transcriptional regulation of both the LS and SS subunits of
Proceedings of the 8th Starch Digestion Consortium Workshop, December 21-22, 2011, Baylor, Texas.
Journal of pediatric gastroenterology and nutrition, 55 Suppl 2, S1-47 (2013-04-27)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service