Skip to Content
Merck
All Photos(1)

Documents

S1100000

Spiramycin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Formacidine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

spiramycin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CO[C@H]1[C@H](O)CC(=O)O[C@H](C)C\C=C\C=C\[C@H](OC2CCC(C(C)O2)N(C)C)[C@H](C)C[C@H](CC=O)[C@@H]1OC3OC(C)C(OC4CC(C)(O)C(O)C(C)O4)C(C3O)N(C)C

InChI

1S/C43H74N2O14/c1-24-21-29(19-20-46)39(59-42-37(49)36(45(9)10)38(27(4)56-42)58-35-23-43(6,51)41(50)28(5)55-35)40(52-11)31(47)22-33(48)53-25(2)15-13-12-14-16-32(24)57-34-18-17-30(44(7)8)26(3)54-34/h12-14,16,20,24-32,34-42,47,49-51H,15,17-19,21-23H2,1-11H3/b13-12+,16-14+/t24-,25-,26?,27?,28?,29+,30?,31-,32+,34?,35?,36?,37?,38?,39+,40+,41?,42?,43?/m1/s1

InChI key

ACTOXUHEUCPTEW-JMRHEKERSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Spiramycin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Severe congenital toxoplasmosis due to a Toxoplasma gondii strain with an atypical genotype: case report and review.
Laurence Delhaes et al.
Prenatal diagnosis, 30(9), 902-905 (2010-06-29)
Ming-juan Wang et al.
Journal of chromatography. A, 1217(42), 6531-6544 (2010-09-17)
Investigation of acetylspiramycin (ASPM) and its related substances was carried out using a reversed-phase liquid chromatography/tandem mass spectrometry method. The identification of impurities in the ASPM complex was performed with a quadrupole ion trap mass spectrometer, with an electrospray ionization
B Julliac et al.
International journal of obstetric anesthesia, 19(3), 331-332 (2010-07-16)
Significant fetal bradycardia occurred when a parturient receiving labor epidural analgesia experienced generalized numbness and tingling, a metallic taste and hot flushes. An emergent cesarean delivery under general anesthesia was performed with favorable outcomes for the mother and baby. The
Yonca Acikgoz et al.
Pediatric transplantation, 16(4), E115-E119 (2011-02-16)
Cryptosporidium is an intracellular protozoan parasite that causes gastroenteritis in human. In immunocompromised individuals, cryptosporidium causes far more serious disease. There is no effective specific therapy for cryptosporidiosis, and spontaneous recovery is the rule in healthy individuals. However, immunocompromised patients
Alain Berrébi et al.
American journal of obstetrics and gynecology, 203(6), 552-552 (2010-07-17)
Maternal toxoplasmosis infection acquired during pregnancy carries significant risk of fetal damage. We aimed to assess the long-term outcome of children and young adults with congenital toxoplasmosis diagnosed and treated in utero. This was a 20 year prospective study (1985-2005).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service