Skip to Content
Merck
All Photos(1)

Key Documents

PHR1955

Supelco

Mepivacaine hydrochloride

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Mepivacaine hydrochloride, 1-Methyl-2′,6′-pipecoloxylidine hydrochloride, N-(2,6-Dimethylphenyl)-1-methyl-2-piperidinecarboxamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22N2O · HCl
CAS Number:
Molecular Weight:
282.81
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 699
traceable to Ph. Eur. M0370000
traceable to USP 1387002

API family

mepivacaine

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

application(s)

pharmaceutical

format

neat

storage temp.

2-30°C

SMILES string

CN1CCCCC1C(NC2=C(C)C=CC=C2C)=O.Cl

InChI

1S/C15H22N2O.ClH/c1-11-7-6-8-12(2)14(11)16-15(18)13-9-4-5-10-17(13)3;/h6-8,13H,4-5,9-10H2,1-3H3,(H,16,18);1H

InChI key

RETIMRUQNCDCQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Mepivacaine hydrochloride is a local anesthetic that is related chemically and pharmacologically to the amide-type local anesthetics. It is often administered via local infiltration, peripheral nerve block, etc.

Application

Mepivacaine hydrochloride may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Local anesthetic. Reversibly blocks transient Na+ inward current, as well as the steady-state K+ outward current. Blocks tandem pore (TASK) and Kv1.5, potassium channels in model systems.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB1955 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mepivacaine hydrochloride
USP42-NF37
United States Pharmacopeia/National Formulary, 38(6), 2743-2743 (2018)
The Estimation of Mepivacain Hydrochloride Injection Liquid
Hazarathaiah C
International Journal of Engineering and Techniques null
Determination of local anaesthetics and their impurities in pharmaceutical preparations using HPLC method with amperometric detection
Fijalek Z, et al.
Journal of Pharmaceutical and Biomedical Analysis, 37(5), 913-918 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service