Skip to Content
Merck
All Photos(1)

Key Documents

PHR1492

Supelco

Phenytoin sodium

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

5,5-Diphenylhydantoin sodium salt, 5,5-Diphenyl-2,4-imidazolidinedione, Phenytoin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C15H11N2O2Na
CAS Number:
Molecular Weight:
274.25
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. P1300000
traceable to USP 1535507

API family

phenytoin

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

[Na]N1C(=O)NC(C1=O)(c2ccccc2)c3ccccc3

InChI

1S/C15H12N2O2.Na/c18-13-15(17-14(19)16-13,11-7-3-1-4-8-11)12-9-5-2-6-10-12;/h1-10H,(H2,16,17,18,19);/q;+1/p-1

InChI key

FJPYVLNWWICYDW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Phenytoin sodium may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by various chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Reduces incidence of grand mal seizures; appears to stabilize excitable membranes perhaps through effects on Na+, K+, and Ca2+ channels.
Reduces incidence of grand mal seizures; appears to stabilize excitable membranes.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA3040 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Differential derivative spectrophotometric determination of phenobarbitone and phenytoin sodium in combined tablet preparations
Prasad CVN, et al.
Talanta, 44(5), 917-922 (1997)
Zin-Juan Klaft et al.
Neuropharmacology, 176, 108213-108213 (2020-07-03)
Status epilepticus (SE) is a neurological emergency characterized by continuous seizure activity lasting longer than 5 min, often with no recovery between seizures (Trinka et al., 2015). SE is refractory to benzodiazepine and second-line treatments in about 30% cases. Novel treatment
Method Development and Validation of Phenytoin Sodium in bulk and its Pharmaceutical dosage form by RP-HPLC method
Varaprasad A, et al.
International Journal of Biological & Pharmaceutical Research, 3(5), 126-129 (2012)
Maria B Rabaglino et al.
BMC genomics, 15, 1001-1001 (2014-11-21)
Major changes in gene expression occur in the fetal brain to modulate the function of this organ postnatally. Thus, factors can alter the genomics of the fetal brain, predisposing to neurological disorders later in life. We hypothesized that the physiological
Misty Good et al.
American journal of physiology. Gastrointestinal and liver physiology, 306(11), G1021-G1032 (2014-04-20)
Necrotizing enterocolitis (NEC) is the leading cause of death from gastrointestinal disease in premature infants and develops partly from an exaggerated intestinal epithelial immune response to indigenous microbes. There has been interest in administering probiotic bacteria to reduce NEC severity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service