Skip to Content
Merck
All Photos(1)

Key Documents

PHR1437

Supelco

Temozolomide

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Temozolomide, 3,4-Dihydro-3-methyl-4-oxoimidazo[5,1-d]-1,2,3,5-tetrazine-8-carboxamide, 3-Methyl-4-oxo-8-imidazolo[5,1-d][1,2,3,5]tetrazinecarboxamide, 4-Methyl-5-oxo-2,3,4,6,8-pentazabicyclo[4.3.0]nona-2,7,9-triene-9-carboxamide, 8-Carbamoyl-3-methylimidazo[5,1-d]-1,2,3,5-tetrazin-4(3H)-one, NSC 362856

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N6O2
CAS Number:
Molecular Weight:
194.15
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1643543

API family

temozolomide

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CN1N=Nc2c(ncn2C1=O)C(N)=O

InChI

1S/C6H6N6O2/c1-11-6(14)12-2-8-3(4(7)13)5(12)9-10-11/h2H,1H3,(H2,7,13)

InChI key

BPEGJWRSRHCHSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to in-house working standards.
Temozolomide is an alkylating agent of the imidazotetrazine group. It is a chemotherapy drug, used in the treatment of anaplastic astrocytoma, glioblastoma multiforme and malignant metastatic melanoma, which works by arresting cancer cell growth.

Application

Temozolomide may be used as a pharmaceutical reference standard for the determination of the analyte in drug dosage forms by reverse phase liquid chromatography and UV spectrophotometry.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA2745 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of UV method of temozolomide in bulk and capsule formulation.
Razak AA, et al.
International Journal of Pharmaceutical Sciences and Research, 4(4), 1419-1423 (2013)
Design of experiment based validated stability indicating RP-HPLC method of temozolomide in bulk and pharmaceutical dosage forms.
Khan A, et al.
Beni-Suef University Journal of Basic and Applied Sciences, 5(4), 402-408 (2016)
Development and Validation of Simultaneous Detrmination of Anastrozole and Temozolomide in Pharmaceutical Dosage Forms.
Daphal VN, et al.
International Journal of Theoretical & Applied Sciences, 4(2), 48-55 (2012)
Reshma Rangwala et al.
Autophagy, 10(8), 1369-1379 (2014-07-06)
Blocking autophagy with hydroxychloroquine (HCQ) augments cell death associated with alkylating chemotherapy in preclinical models. This phase I study evaluated the maximum tolerated dose (MTD), safety, preliminary activity, pharmacokinetics, and pharmacodynamics of HCQ in combination with dose-intense temozolomide (TMZ) in
Kelly Burrell et al.
Cancer research, 74(14), 3727-3739 (2014-05-14)
Glioblastoma multiforme (GBM) is characterized by a pathogenic vasculature that drives aggressive local invasion. Recent work suggests that GBM cells recruit bone marrow-derived progenitor cells (BMDC) to facilitate recurrence after radiotherapy, but how this may be achieved is unclear. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service