Skip to Content
Merck
All Photos(1)

Key Documents

PHR1430

Supelco

Dihydroxyacetone

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Dihydroxyacetone, 1,3-Dihydroxy-2-propanone, 1,3-Dihydroxyacetone, DHA, Glycerone, NSC 24343

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O3
CAS Number:
Molecular Weight:
90.08
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1204102

API family

dihydroxyacetone

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

O1C(COC(C1)(O)CO)(O)CO

InChI

1S/C6H12O6/c7-1-5(9)3-12-6(10,2-8)4-11-5/h7-10H,1-4H2

InChI key

KEQUNHIAUQQPAC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dihydroxyacetone (DHA) is a browning ingredient widely used in cosmetics such as sunless tanning formulations. It participates in a chemical staining reaction called Milliard reaction in which it reacts with the amino groups of proteins to result in a mixture of high molecular weight pigments.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Dihydroxyacetone may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by liquid chromatography (LC) technique.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC0574 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dihydroxyacetone
USP42-NF37: United States Pharmacopeia and National Formulary
United States Pharmacopeia/National Formulary, 1366-1366 (2018)
Influence of hydration on dihydroxyacetone-induced pigmentation of stratum corneum
Nguyen BC and Kochevar IE
The Journal of Investigative Dermatology, 120(4), 655-661 (2003)
Dihydroxyacetone, the active browning ingredient in sunless tanning lotions, induces DNA damage, cell-cycle block and apoptosis in cultured HaCaT keratinocytes
Petersen AB, et al.
Mutation Research, 560(2), 173-186 (2004)
Megan A Rees et al.
Journal of proteome research, 14(1), 120-132 (2014-10-21)
Interactions between a host and a bacterial pathogen are mediated by cross-talk between molecules present on, or secreted by, pathogens and host binding-molecules. Identifying proteins involved at this interface would provide substantial insights into this interaction. Although numerous studies have
C Flavia Massaro et al.
Journal of agricultural and food chemistry, 62(50), 12209-12217 (2014-11-26)
Australian stingless bee honeys have been shown to exert antioxidant and in vitro antimicrobial properties; however their bioactive factors remained unidentified. This study investigated the antibacterial properties of phenolic extracts from Tetragonula carbonaria honeys. Honeys were harvested from beehives in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service