Skip to Content
Merck
All Photos(1)

Key Documents

PHR1381

Supelco

Aspartame

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Asp-Phe methyl ester, N-(L-α-Aspartyl)-L-phenylalanine methyl ester, Asp-Phe-OMe, Aspartame

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH2CH(NH2)CONHCH(CH2C6H5)COOCH3
CAS Number:
Molecular Weight:
294.30
Beilstein:
2223850
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. A1320000
traceable to USP 1043706

API family

aspartame

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

COC(=O)[C@H](Cc1ccccc1)NC(=O)[C@@H](N)CC(O)=O

InChI

1S/C14H18N2O5/c1-21-14(20)11(7-9-5-3-2-4-6-9)16-13(19)10(15)8-12(17)18/h2-6,10-11H,7-8,15H2,1H3,(H,16,19)(H,17,18)/t10-,11-/m0/s1

InChI key

IAOZJIPTCAWIRG-QWRGUYRKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Aspartame is a low-calorie nutritive sugar substitute used in table-top sweeteners, chewing gums, soft drinks, etc.

Application

Aspartame may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by high performance liquid chromatography.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB9987 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HPLC analysis of aspartame and saccharin in pharmaceutical and dietary formulations.
Di Pietra AM, et al.
Chromatographia, 30(3-4), 215-219 (1990)
B A Magnuson et al.
Critical reviews in toxicology, 37(8), 629-727 (2007-09-11)
Aspartame is a methyl ester of a dipeptide used as a synthetic nonnutritive sweetener in over 90 countries worldwide in over 6000 products. The purpose of this investigation was to review the scientific literature on the absorption and metabolism, the
Karol Rycerz et al.
Folia neuropathologica, 51(1), 10-17 (2013-04-05)
Aspartame, a widespread sweetener used in many food products, is considered as a highly hazardous compound. Aspartame was discovered in 1965 and raises a lot of controversy up to date. Astrocytes are glial cells, the presence and functions of which
P Humphries et al.
European journal of clinical nutrition, 62(4), 451-462 (2007-08-09)
The use of the artificial sweetener, aspartame, has long been contemplated and studied by various researchers, and people are concerned about its negative effects. Aspartame is composed of phenylalanine (50%), aspartic acid (40%) and methanol (10%). Phenylalanine plays an important
Harriett H Butchko et al.
Regulatory toxicology and pharmacology : RTP, 35(2 Pt 2), S1-93 (2002-08-16)
Over 20 years have elapsed since aspartame was approved by regulatory agencies as a sweetener and flavor enhancer. The safety of aspartame and its metabolic constituents was established through extensive toxicology studies in laboratory animals, using much greater doses than

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service