Skip to Content
Merck
All Photos(1)

Key Documents

PHR1196

Supelco

Nitrofurazone

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

5-Nitro-2-furaldehyde semicarbazone, NZ, Nitrofural, 5-Nitro-2-furfural semicarbazone, Furacin, NFZ, Nitrofurazone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N4O4
CAS Number:
Molecular Weight:
198.14
Beilstein:
86403
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. N0950000
traceable to USP 1465004

API family

nitrofurazone

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

242-244 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

NC(=O)N\N=C\c1ccc(o1)[N+]([O-])=O

InChI

1S/C6H6N4O4/c7-6(11)9-8-3-4-1-2-5(14-4)10(12)13/h1-3H,(H3,7,9,11)/b8-3+

InChI key

IAIWVQXQOWNYOU-FPYGCLRLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Nitrofurazone may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by DC polarography, chromatography and spectrophotometric techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC0296 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Muta. 2 - Repr. 2 - Skin Sens. 1 - STOT RE 2

Target Organs

male reproductive organs

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical reduction of nitrofurazone and its determination in pharmaceutical dosage forms by Dc polarography
Mishra AK and Gode KD
Analyst, 110(11), 1373-1376 (1985)
Inclusion compounds in pharmaceutical analysis II: Assay of a cream following chemical dehydration
Hays SE and Grady LT
Journal of Pharmaceutical Sciences, 60(2), 295-298 (1971)
Teresa del Castillo et al.
ChemMedChem, 9(2), 383-389 (2013-12-18)
Cyclodextrins have been conjugated to target various receptors and have also been functionalized with carbohydrates for targeting specific organs. However, this approach is based on a rigid design that implies the ad hoc synthesis of each cyclodextrin-targeting agent conjugate. We
Hamid R Moghimi et al.
International journal of pharmaceutics, 401(1-2), 47-50 (2010-09-22)
Microwaves (MW), a part of the electromagnetic spectrum at 0.3-300GHz, affect human body in different ways through its thermal and athermal effects, including fluidization of cell membranes and liquid crystalline systems. Due to presence of such structures in skin barrier
Arnaud Gutierrez et al.
Nucleic acids research, 39(10), 4192-4201 (2011-02-09)
Escherichia coli SOS functions constitute a multifaceted response to DNA damage. We undertook to study the role of yafP, a SOS gene with unknown function. yafP is part of an operon also containing the dinB gene coding for DNA Polymerase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service