Skip to Content
Merck
All Photos(1)

Key Documents

PHR1143

Supelco

Lisinopril

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

(S)-1-[N2-(1-carboxy-3-phenylpropyl)-lysyl-proline dihydrate, MK-521

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H31O5N3 · 2H2O
CAS Number:
Molecular Weight:
441.52
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 695
traceable to Ph. Eur. L0702000
traceable to USP 1368609

API family

lisinopril

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

[H]O[H].[H]O[H].NCCCC[C@H](N[C@@H](CCc1ccccc1)C(O)=O)C(=O)N2CCC[C@H]2C(O)=O

InChI

1S/C21H31N3O5.2H2O/c22-13-5-4-9-16(19(25)24-14-6-10-18(24)21(28)29)23-17(20(26)27)12-11-15-7-2-1-3-8-15;;/h1-3,7-8,16-18,23H,4-6,9-14,22H2,(H,26,27)(H,28,29);2*1H2/t16-,17-,18-;;/m0../s1

InChI key

CZRQXSDBMCMPNJ-ZUIPZQNBSA-N

Gene Information

human ... ACE(1636)

Looking for similar products? Visit Product Comparison Guide

General description

Lisinopril is a lysine derivative of enalaprilat and belongs to the group of angiotensin converting enzyme (ACE) inhibitors. It is widely used as an orally active medication in the treatment of high blood pressure and may be used to treat congestive heart failure.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Lisinopril may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using kinetic spectrophotometric technique.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Angiotensin converting enzyme (ACE) inhibitor.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Values of analytes vary lot to lot.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB3302 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1A - STOT RE 2

Target Organs

Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jason V Baker et al.
PloS one, 7(10), e46894-e46894 (2012-10-20)
Treatments that reduce inflammation and cardiovascular disease (CVD) risk among individuals with HIV infection receiving effective antiretroviral therapy (ART) are needed. We conducted a 2 × 2 factorial feasibility study of lisinopril (L) (10 mg daily) vs L-placebo in combination
Determination of lisinopril in pharmaceuticals by a kinetic spectrophotometric method
Cakar M and Popovic G
J. Serb. Chem. Soc., 77(10), 1437-1442 (2012)
Lisinopril
Lancaster GS and Todd AP
Drugs, 35(6), 646-669 (1988)
Saleh Yazdani et al.
PloS one, 7(11), e50209-e50209 (2012-11-29)
Proteinuria is an important cause of progressive tubulo-interstitial damage. Whether proteinuria could trigger a renal lymphangiogenic response has not been established. Moreover, the temporal relationship between development of fibrosis, inflammation and lymphangiogenesis in chronic progressive kidney disease is not clear
K L Goa et al.
Drugs, 53(6), 1081-1105 (1997-06-01)
Lisinopril, like other ACE inhibitors, lowers blood pressure and preserves renal function in hypertensive patients with non-insulin-dependent or insulin-dependent diabetes mellitus (NIDDM or IDDM) and early or overt nephropathy, without adversely affecting glycaemic control or lipid profiles. On available evidence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service