Skip to Content
Merck
All Photos(1)

Documents

PHR1135

Supelco

Myristyl Alcohol

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

1-Tetradecanol, Myristyl alcohol, Tetradecyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13OH
CAS Number:
Molecular Weight:
214.39
Beilstein:
1742652
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1449008

vapor density

7.4 (vs air)

API family

myristyl alcohol

CofA

current certificate can be downloaded

autoignition temp.

489 °F

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

289 °C (lit.)

mp

35-39 °C (lit.)

density

0.823 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CCCCCCCCCCCCCCO

InChI

1S/C14H30O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15/h15H,2-14H2,1H3

InChI key

HLZKNKRTKFSKGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Myristyl Alcohol was used for filling the hollow interiors of gold nanocages in the fabrication of new theranostic system having unique feature of photoacoustic imaging. It was used in fabrication of temperature-regulated drug release system based on phase-change materials.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Values of analytes vary lot to lot.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA7155 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

298.4 °F - closed cup

Flash Point(C)

148 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patch testing with myristyl alcohol.
Johannes Geier et al.
Contact dermatitis, 55(6), 366-367 (2006-11-15)
M Lafleur et al.
Biochemistry, 29(36), 8325-8333 (1990-09-11)
The orientational order profile has been determined by using deuterium nuclear magnetic resonance (2H NMR) for POPE in the lamellar liquid-crystalline (L alpha) and the hexagonal (HII) phases and is shown to be sensitive to the symmetry of the lipid
L Kato et al.
International journal of leprosy and other mycobacterial diseases : official organ of the International Leprosy Association, 62(1), 75-88 (1994-03-01)
Host-grown Mycobacterium leprae cell suspensions oxidized water-soluble complexes of palmitic acid, myristic acid, cetyl alcohol, and myristyl alcohol prepared with randomly methylated-beta-cyclodextrin as host molecules. Gas chromatography analysis showed that the water-soluble complexes retained their chemical structure following sterilization in
Antonio Ortiz et al.
Journal of chemical ecology, 30(5), 991-1000 (2004-07-28)
The potential for pheromone-based mating disruption of the olive pyralid moth (OPM), Euzophera pinguis, in olive groves was investigated during the second flight period in small-plot trials in 2002. The female of this species emits a blend of (9Z,12E)-tetradecadien-1-ol and
Yijun Du et al.
Virus research, 147(2), 294-299 (2009-12-03)
The small envelope (E) protein of porcine reproductive and respiratory syndrome virus (PRRSV) is known to possess the properties of an ion-channel protein, and in the present study we show that the PRRSV E protein is N-terminal myristoylated. The PRRSV

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service