Skip to Content
Merck
All Photos(1)

Key Documents

PHR1130

Supelco

Potassium gluconate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Potassium D-gluconate, 2,3,4,5,6-Pentahydroxycaproic acid potassium salt, D-Gluconic acid potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11KO7
CAS Number:
Molecular Weight:
234.25
Beilstein:
3919448
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
E Number:
E577
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1550001

API family

potassium gluconate

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

183 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

[K+].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O

InChI

1S/C6H12O7.K/c7-1-2(8)3(9)4(10)5(11)6(12)13;/h2-5,7-11H,1H2,(H,12,13);/q;+1/p-1/t2-,3-,4+,5-;/m1./s1

InChI key

HLCFGWHYROZGBI-JJKGCWMISA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium Gluconate is a potassium salt of gluconic acid. It is a commonly administered nutrient supplement form of potassium.[1]
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Potassium Gluconate may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography.[2]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA7151in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potassium Gluconate
USP42-NF37: United States Pharmacopeia and National Formulary
United States Pharmacopeia/National Formulary, 41(6), 3590-3590 (2018)
Herb, nutrient, and drug interactions: clinical implications and therapeutic strategies.
Stargrove MB, et al.
Herb, Nutrient, and Drug Interactions: Clinical Implications and Therapeutic Strategies (2008)
Ubaid H Shah et al.
Clinical nutrition (Edinburgh, Scotland), 32(2), 193-199 (2012-09-18)
Acute lower respiratory infections are the most frequent illnesses globally in children less than 5 years old. The aim of this randomized double blind controlled trial is to assess the effectiveness of zinc gluconate supplementation for 2 months period compared
Ze-Ping Hu et al.
Journal of proteome research, 11(12), 5903-5913 (2012-10-20)
Identification of molecular mechanisms underlying early stage Alzheimer's disease (AD) is important for the development of new therapies against and diagnosis of AD. In this study, nontargeted metabonomics of TASTPM transgenic AD mice was performed. The metabolic profiles of both
Hossam E Emam et al.
Carbohydrate polymers, 90(3), 1345-1352 (2012-09-04)
Copper containing cellulose material is of growing interest, e.g. offering alternative in the field of antimicrobials. Solutions of copper d-gluconate complexes (Cu(2+)-DGL) were used to introduce copper ions into a swollen cellulosic matrix. A ligand exchange mechanism forms the chemical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service