Skip to Content
Merck
All Photos(4)

Documents

P5338

Sigma-Aldrich

3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4′,4′′-disulfonic acid sodium salt

Powder

Synonym(s):

5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4′,4″-disulfonic acid sodium salt, PDT disulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H13N4NaO6S2
CAS Number:
Molecular Weight:
492.46
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4′,4′′-disulfonic acid sodium salt, BioXtra

product line

BioXtra

Quality Level

form

powder

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

mp

≥300 °C

anion traces

chloride (Cl-): ≤0.1%

cation traces

Al: ≤0.001%
Ca: ≤0.01%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.001%
Pb: ≤0.001%
Zn: ≤0.0005%

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].OS(=O)(=O)c1ccc(cc1)-c2nnc(nc2-c3ccc(cc3)S([O-])(=O)=O)-c4ccccn4

InChI

1S/C20H14N4O6S2.Na/c25-31(26,27)15-8-4-13(5-9-15)18-19(14-6-10-16(11-7-14)32(28,29)30)23-24-20(22-18)17-3-1-2-12-21-17;/h1-12H,(H,25,26,27)(H,28,29,30);/q;+1/p-1

InChI key

ZGVNYCXXBQPDPQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4′,4′′-disulfonic acid sodium salt is also known as ferrozine.

Application

3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4′,4′′-disulfonic acid sodium salt (ABTS) has been used in the ferene-s assay for the accurate determination of intracellular iron from iron oxide nanoparticles.
It has also been used in ABTS radical scavenging assay to determine the antioxidant activity of honey samples.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William W Bennett et al.
Talanta, 88, 490-495 (2012-01-24)
The recently developed colorimetric diffusive equilibration in thin films (DET) technique for the in situ, high-resolution measurement of iron(II) in marine sediments is optimized to allow measurement of the higher iron concentrations typical of freshwater sediment porewaters. Computer imaging densitometry
Aya Usami et al.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine, 24(6), 1217-1224 (2011-07-07)
Bovine milk α-casein was identified as a ferritin-binding protein, and ferritin is known to be a heme-binding protein. In this study, we found that the binding of α-casein to bovine spleen ferritin in vitro was blocked by hemin, but not
Mitsuo Yamamoto et al.
Bioresource technology, 101(12), 4456-4460 (2010-02-19)
In evaluating the quality of composts that include humic substances (HSs), the binding of iron(II) (conditional binding constants and binding capacities) are an important criteria. To determine the binding ability, it is necessary to determine the concentrations of free Fe(2+)
James R Reed et al.
Drug metabolism and disposition: the biological fate of chemicals, 38(11), 2060-2066 (2010-08-04)
Heme oxygenase 1 (HO-1) uses molecular oxygen and electrons from NADPH cytochrome P450 reductase to convert heme to CO, ferrous iron, and biliverdin (BV). Enzymatic studies with the purified 30-kDa form of HO-1 routinely use a coupled assay containing biliverdin
Wance J J Firdaus et al.
The FEBS journal, 273(13), 3076-3093 (2006-07-05)
We recently reported that the transient expression of polyglutamine tracts of various size in exon 1 of the huntingtin polypeptide (httEx1) generated abnormally high levels of intracellular reactive oxygen species that directly contributed to cell death. Here, we compared the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service