Skip to Content
Merck
All Photos(5)

Key Documents

P4758

Sigma-Aldrich

Phenol Red sodium salt

Powder

Synonym(s):

Phenolsulfonephthalein sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H13NaO5S
CAS Number:
Molecular Weight:
376.36
Beilstein:
6260026
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Phenol Red sodium salt, pH indicator dye

Quality Level

form

powder

technique(s)

titration: suitable

color

red to brown

visual transition interval

6.8-8.2, yellow to red

mp

285 °C (dec.) (lit.)

solubility

water: 1 mg/mL, opaque, dark red

εmax

31.62 at 423 nm in 0.1 M phosphate buffer

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].Oc1ccc(cc1)C2(OS(=O)(=O)c3ccccc23)c4ccc([O-])cc4

InChI

1S/C19H14O5S.Na/c20-15-9-5-13(6-10-15)19(14-7-11-16(21)12-8-14)17-3-1-2-4-18(17)25(22,23)24-19;/h1-12,20-21H;/q;+1/p-1

InChI key

HKHYOKBQJILTEI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Phenol Red sodium salt is widely used as a pH indicator. At a pH of 6.4 and below, phenol red solution will have a yellow color. At a pH of 8.2 and above, it will have a red color. It is commonly used in culture media to detect neutral to acidic pH changes.

Application

Phenol Red sodium salt has been used as an indicator during single-guide RNA microinjection into one-cell stage zebrafish embryos.
Suitable for use as a pH indicator.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Textbook of Assisted Reproduction for Scientists in Reproductive Technology (2009)
Elmer W. Koneman
Koneman's Color Atlas and Textbook of Diagnostic Microbiology (2006)
BATCH-GE: Batch analysis of Next-Generation Sequencing data for genome editing assessment
Annekatrien Boel
Scientific Reports, 6 (2016)
Connie R. Mahon
Textbook of Diagnostic Microbiology (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service