Skip to Content
Merck
All Photos(2)

Key Documents

P4030

Sigma-Aldrich

Phloxine B

Dye content, ≥80%, certified by the Biological Stain Commission, powder

Synonym(s):

2′,4′,5′,7′-Tetrabromo-4,5,6,7-tetrachlorofluorescein disodium salt, Acid Red 92, Cyanosine, D&C;Red No. 28, Eosin 10B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H2Br4Cl4Na2O5
CAS Number:
Molecular Weight:
829.63
Colour Index Number:
45410
Beilstein:
3887100
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Phloxine B, Dye content ≥80 %, certified by the Biological Stain Commission

grade

certified by the Biological Stain Commission

Quality Level

form

powder

composition

Dye content, ≥80%

color

dark red

solubility

H2O: 1 mg/mL

εmax

1150-1300 at 546-550 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].[O-]c1c(Br)cc2c(Oc3c(Br)c([O-])c(Br)cc3C24OC(=O)c5c(Cl)c(Cl)c(Cl)c(Cl)c45)c1Br

InChI

1S/C20H4Br4Cl4O5.2Na/c21-5-1-3-17(9(23)15(5)29)32-18-4(2-6(22)16(30)10(18)24)20(3)8-7(19(31)33-20)11(25)13(27)14(28)12(8)26;;/h1-2,29-30H;;/q;2*+1/p-2

InChI key

OOYIOIOOWUGAHD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Phloxine B is part of the xanthylium class of dyes. The stain is mostly used to determine cell ploidy and temperature sensitivity of Schizosaccharomyces pombe. Solid media is colored a vivid pink.

Application

Phloxine B has been used as a component of yeast extract peptone dextrose (YEPD) to grow Schizosaccharomyces pombe.

Suitability

Certified for use in Mallory′s phloxine-methylene blue stain; Kreyberg′s method for keratin and mucus.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Advances in Food and Nutrition Research, 61 (2010)
The Small Subunit Processome Is Required for Cell Cycle Progression at G1
Kara A. Bernstein and Susan J. Baserga
Molecular Biology of the Cell (2004)
Basic methods for fission yeast
Susan L
Yeast (2006)
Ellyn E Ritchie et al.
Chemosphere, 90(7), 2129-2135 (2012-12-06)
Soil eco-toxicity testing was conducted in support of Canada's Chemical Management Plan (CMP) to fill data gaps for organic chemicals known to primarily partition to soil, and of which the persistence and inherent toxicity are uncertain. Two compounds representative of
Ander M de Lecea et al.
Rapid communications in mass spectrometry : RCM, 25(13), 1853-1861 (2011-06-04)
Stable isotope measurements are an important tool for ecosystem trophic linkage studies. Ideally, fresh samples should be used for isotopic analysis, but in many cases organisms must be preserved and analysed later. In some cases dyes must be used to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service