Skip to Content
Merck
All Photos(1)

Key Documents

P3800000

Proxyphylline

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

7-(β-Hydroxypropyl)theophylline, (1,3-Dimethyl-7-[2-hydroxypropyl]-2,6-dioxopurine), Proxyphylline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N4O3
CAS Number:
Molecular Weight:
238.24
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

proxyphylline

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(O)Cn1cnc2N(C)C(=O)N(C)C(=O)c12

InChI

1S/C10H14N4O3/c1-6(15)4-14-5-11-8-7(14)9(16)13(3)10(17)12(8)2/h5-6,15H,4H2,1-3H3

InChI key

KYHQZNGJUGFTGR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Proxyphylline EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Magnussen et al.
Deutsche medizinische Wochenschrift (1946), 111(1), 15-19 (1986-01-03)
In study I, carried out on 9 asthmatics to investigate whether the bronchodilator and protective effect of intravenous theophylline, dissolved in ethylene diamine (T.E.) or proxyphylline and diprophylline (T.P.D.), showed different dose-effect relationships, 100 mg theophylline with T.E., T.P.D. or
Linfeng Wu et al.
International journal of pharmaceutics, 349(1-2), 1-10 (2007-09-11)
Delayed release systems find applications in chronotherapeutics and colon-specific delivery. They have also been considered suitable carriers for the oral delivery of peptides and proteins. In prior work, our research group has reported surface crosslinking as an effective technique to
R Duclos et al.
International journal of pharmaceutics, 182(2), 145-154 (1999-05-26)
Three polyol behenates with similar melting points (MP) and different hydrophilic-lipophilic balances (HLB) were studied (MP/HLB: 70/02, 63/05 and 57/13). After melting at MP+30 degrees C, the rheological behaviour of behenates was determined by adjustment of the rheograms to the
H Magnussen et al.
Chest, 90(5), 722-725 (1986-11-01)
To compare the acute bronchodilator effect of increasing doses of intravenous theophylline and inhaled beta adrenergic agonists, we administered intravenous theophylline dissolved in ethylenediamine or proxyphylline and diprophylline or placebo in a double blind fashion to nine asthmatics on three
D Agbaba et al.
Biomedical chromatography : BMC, 6(3), 141-142 (1992-05-01)
The content and dissolution rate of theophylline, diprophylline and proxyphylline from a sustained release formulation were determined by UV in situ densitometry. After separation the chromatographic zones corresponding to the spots of theophylline, diprophylline and proxyphylline on the high performance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service