Skip to Content
Merck
All Photos(1)

Key Documents

P1900000

Piperazine adipate

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20N2O4
CAS Number:
Molecular Weight:
232.28
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

piperazine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

C1CNCCN1.OC(=O)CCCCC(O)=O

InChI

1S/C6H10O4.C4H10N2/c7-5(8)3-1-2-4-6(9)10;1-2-6-4-3-5-1/h1-4H2,(H,7,8)(H,9,10);5-6H,1-4H2

InChI key

BVEGEKOBSPXUJS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Piperazine adipate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

>194.0 °F - closed cup

Flash Point(C)

> 90 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Experience with the prevention of enterobiasis in children from preschool institutions of the Zhitomir district].
V A Pomazan et al.
Pediatriia, (7)(7), 82-83 (1990-01-01)
M Grzywacz
Polish journal of pharmacology and pharmacy, 38(4), 335-341 (1986-07-01)
The whole integumentomuscular sac of the worm Ascaris lumbricoides suis L. and pouches isolated from it seemed to be a good model for investigation of the transmembrane transport of chemical substances across tissue barriers.
Iu I Kozin et al.
Problemy tuberkuleza, (6)(6), 29-32 (1991-01-01)
Findings of a complex immunologic examination of 138 patients with active cavernous tuberculosis of the kidneys concurrent with secondary immunodeficiency are presented. Biostimulators and immunomodulating drugs were administered as conservative treatment in this case. Changes in cellular and humoral immunity
[Effectiveness and influence of piperazine adipinate on the immunological indices of furunculosis patients].
L N Savitskaia et al.
Vestnik dermatologii i venerologii, (9)(9), 9-12 (1985-09-01)
M A Fisher et al.
Veterinary parasitology, 55(1-2), 87-92 (1994-10-01)
The control of Toxocara canis was investigated in naturally infected unweaned puppies. Anthelmintic treatments were administered to three litters of pups at 2, 4 and 6 weeks of age. When either a new combination anthelmintic containing febantel, pyrantel embonate and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service