Skip to Content
Merck
All Photos(1)

Key Documents

NIST1991

Mix coal tar/petroleum extract in methylene chloride

NIST® SRM® 1991

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 1.2 mL

manufacturer/tradename

NIST®

technique(s)

GC/MS: suitable

application(s)

industrial hygiene

format

matrix material

General description

Other Notes

Certified for the analytes listed below.
Naphthalene, 1-Methylnaphthalene, 2-Methylnaphthalene, Biphenyl, Dibenzothiophene, Phenanthrene, Anthracene, 1-Methylphenanthrene, 3-Methylphenanthrene, 9-Methylphenanthrene, 2-Methylanthracene, 4H-Cyclopenta[def]phenanthrene, Fluoranthene, Pyrene, Benzo[ghi]fluoranthene, Benzo[c]phenanthrene, Benz[a]anthracene, Chrysene, Benzo[a]fluoranthene, Benzo[b]fluoranthene, Benzo[j]fluoranthene, Benzo[k]fluoranthene, Benzo[e]pyrene, Benzo[a]pyrene, Perylene, Benzo[ghi]perylene, Indeno[1,2,3-cd]pyrene, Dibenz[a,c]anthracene, Dibenz[a,h]anthracene, Benzo[b]chyrsene, Picene

Reference/Informational value is provided for the following analytes.
Selected polycyclic aromatic hydrocarbons (PAHs): Acenaphthene, Fluorene, 1-Methylfluoranthene, 3-Methylfluoranthene, 1-Methylpyrene, 4-Methylpyrene, Cyclopenta[cd]pyrene, Triphenylene, 3-Methylchrysene, Anthanthrene, Dibenz[a,j]anthracene, Coronene, Dibenzo[b,k]fluoranthene, Dibenzo[a,e]pyrene, Dibenzo[a,h]pyrene
Alkylated PAH Groups: C2-naphthalenes, C3-naphthalenes, C4-naphthalenes, C1-fluorenes, C2-fluorenes, C3-fluorenes, C1-dibenzothiophenes, C2-dibenzothiophenes, C3-dibenzothiophenes, C1-phenanthrenes/anthracenes, C2-phenanthrenes/anthracenes, C3-phenanthrenes/anthracenes, C4-phenanthrenes/anthracenes, C1-fluoranthenes/pyrenes, C2-fluoranthenes/pyrenes, C1-benzanthracenes/chrysenes/triphenylenes, C2-benzanthracenes/chrysenes/triphenylenes

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service