Skip to Content
Merck
All Photos(1)

Key Documents

NIST195

Ferrosilicon

NIST® SRM® 195, 75% Si - high-purity grade

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

packaging

pkg of 75 g

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

A single unit of SRM 195 is composed of a bottle consisting of approximately 75 g of pulverized fine powder (< 0.15 mm). For more information, please refer to the SDS and COA.
SRM 195_cert
SRM 195_SDS

Application

Ferrosilicon Standard Reference Material (SRM) is intended for use in the validation of chemical and instrumental methods of analysis.

Features and Benefits

  • Accompanied with certificate documenting NIST-certified mass fraction values, coverage factor (k), and expanded uncertainty for total constituents in ferrosilicon and non-certified values of constituents in ferrosilicon.
  • Information values for constituents are provided and expressed in mass fraction.
  • The NIST certificate includes expiration details, usage, and storage instructions.

Other Notes

Certified for the analytes listed below.
Boron (B), Chromium (Cr), Copper (Cu), Iron (Fe), Manganese (Mn), Nickel (Ni), Silicon (Si)

Reference/Informational value is provided for the following analytes.
Aluminium (Al), Carbon (C), Calcium (Ca), Phosphorus (P), Titanium (Ti), Zirconium (Zr), Cobalt (Co), Molybdinum (Mo), Oxygen (O), Sulfur (S), Tin (Sn)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Can I know the particle size?

    1 answer
    1. 1 unit of NIST195 consists of a bottle containing approximately 75 g of pulverized, fine powder of (< 0.15 mm) particle size.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service