Skip to Content
Merck
All Photos(1)

Documents

NIST1889B

Portland Cement (Blended with Limestone)

NIST®SRM®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

form

powder

packaging

pkg of 5 vials x 5 g

manufacturer/tradename

NIST®

storage temp.

15-25°C

General description

One unit of SRM 1889b contains five vials, with each vials containing ~ 5 g of cement in a foil pouch, which is ground to pass through a sieve of 75 µm (No. 200).


SRM 1889B_cert

SRM 1889B_SDS

Application

The SRM is a Portland cement blended with limestone, intended to evaluate the methods used to analyze the cements and similar matrix materials.

Features and Benefits

  • Accompanied with certificate documenting NIST- certified mass fraction values, and coverage factor for the constituents in Portland cement.
  • The reference and information values of constituents are reported as mass fraction.
  • The certified value is metrologically traceable to the SI unit of mass fraction, expressed as a percentage.
  • The NIST certificate is provided with expiration certificate, use, storage, handling, and maintenance instructions.

Other Notes

Certified for the analytes listed below.
Silicon Dioxide (SiO2), Aluminum Oxide (Al2O3), Ferric Oxide (Fe2O3), Calcium Oxide (CaO), Magnesium Oxide (MgO), Sulfur trioxide (SO3), Sodium Oxide (Na2O), Potassium oxide (K2O), Titanium Dioxide (TiO2), Phosphorus pentoxide (P2O5), Manganese(III) oxide (Mn2O3), Chromium(III) oxide (Cr2O3), Zinc oxide (ZnO), Strontium oxide (SrO), Chlorine (Cl)

Reference/Informational value is provided for the following analytes.
Sulfide sulfur, Insoluble residue, Free CaO, Fluoride (F)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service