Skip to Content
Merck
All Photos(1)

Key Documents

NIST1882A

Calcium aluminate cement

NIST® SRM® 1882a

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 4 x 5 g

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is intended primarily for use in evaluating chemical methods of analysis and in the calibration of instrumental methods for analysis of cements and materials of similar matrix. A unit of SRM 1882a consists of four sealed vials, each containing approximately 5 g of calcium aluminate cement. For more information, please refer to the SDS and the COA.

SRM 1882A_cert

SRM 1882A_SDS

Other Notes

Certified for the analytes listed below.
Silicon dioxide (SiO2), Magnesium oxide (MgO), Aluminium oxide (Al2O3), Sodium oxide (Na2O), Iron(III) oxide (Fe2O3), Potassium oxide (K2O), Calcium oxide (CaO), Titanium dioxide (TiO2)

Reference/Informational value is provided for the following analytes.
Phosphorus pentoxide (P2O5), Strontium oxide (SrO), Zinc oxide (ZnO), Chromium(III) oxide (Cr2O3), Manganese(III) oxide (Mn2O3)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service