Skip to Content
Merck
All Photos(1)

Key Documents

NIST1650B

Diesel particulate matter

NIST® SRM® 1650b

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

packaging

pkg of 200 mg

manufacturer/tradename

NIST®

application(s)

industrial hygiene

format

matrix material

SMILES string

C

InChI

1S/CH4/h1H4

InChI key

VNWKTOKETHGBQD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The particulate material used for the preparation of SRM 1650b was collected from the heat exchangers of a dilution tube facility following 200 engine hours of particulate accumulation. Several direct injection four-cycle diesel engines, operating under a variety of conditions were used to generate this particulatematerial. Therefore, while the sample is not intended to be representative of any particular diesel engine operating under any specific conditions, it should be representative of heavy-duty diesel engine particulate emissions. All of the chemical constituents for which certified and non-certified values are provided, are naturally present in the diesel particulate material. The certified values for the PAHs are based on the agreement of results obtained at NIST from two or more independent analytical methods. SRM 1650b is valid, with the measurement uncertainty specified, provided the SRM is handled and stored in accordance with instructions given under “Instructions forUse” in the certificate.

SRM 1650B_cert

SRM 1650B_SDS

Application

This Standard Reference Material (SRM) is intended for use in evaluating analytical methods for the determination of selected polycyclic aromatic hydrocarbons (PAHs) and nitro-substituted PAHs (nitro-PAHs) in diesel particulate matter and similar matrices.

Features and Benefits

  • A unit of SRM 1650b consists of a bottle containing approximately 200 mg of diesel particulate material
  • Metrological traceability is to the International System of Units (SI) unit of mass expressed as a derived unit of mass fraction
  • Information on product description, disposal, and transportation is available in the accompanying MSDS
  • The enclosed NIST certificate contains details on the expiry date, warning to users, instructions for right storage and use, preparation, and analysis of the RM

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Woo Nam et al.
Journal of medicinal food, 23(6), 575-583 (2020-04-17)
Epidemiological studies have shown that exposure to particulate matter (PM) is associated with adverse health effects. Inhalation of fine particulate matter (PM2.5) is associated with elevated pulmonary diseases. However, the molecular mechanism underlying the initiation of lung inflammation following inhalation

Questions

  1. Is NIST1650B PM2.5?

    1 answer
    1. This item is a product of NIST. While the supplier does not specifically classify the product as PM2.5, the particle size distribution data indicates submicron particles make up the bulk of this product, as shown below. Please see the following link to review the entire Certificate of Analysis:
      https://tsapps.nist.gov/srmext/certificates/1650b.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service