Skip to Content
Merck
All Photos(1)

Key Documents

N4754

Sigma-Aldrich

Nigrosin water soluble

Powder

Synonym(s):

Acid black 2

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Colour Index Number:
50420
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47

product name

Nigrosin water soluble, For use as a biological stain

Quality Level

form

powder

solubility

water: 1 mg/mL, turbid

εmax

~250 at 575 nm in 50% ethanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Nigrosin is an acidic dye used as a negative stain for visualization of the capsule in bacteriological material.

Application

Nigrosin has the following applications:
  • to stain live sperm
  • as a viable cell staining agent especially in stem cell biology
  • as a negative stain in place of India ink for spirochetes, bacteria, protozoa, and fungi
  • in Dorner′s spore stain.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Experiments in Microbiology, Plant Pathology and Biotechnology (2003)
Broadband UV-Vis optical property measurement in layered turbid media.
Wang Q
SPIE Medical Imaging. International Society for Optics and Photonics, 2014 (2011)
Graff's Textbook of Routine Urinalysis and Body Fluids (2010)
Quanzeng Wang et al.
Biomedical optics express, 3(6), 1226-1240 (2012-06-29)
The ability to accurately measure layered biological tissue optical properties (OPs) may improve understanding of spectroscopic device performance and facilitate early cancer detection. Towards these goals, we have performed theoretical and experimental evaluations of an approach for broadband measurement of
Jesse Schallek et al.
Investigative ophthalmology & visual science, 52(3), 1325-1335 (2010-11-06)
To examine the extent to which neurovascular coupling contributes to stimulus-evoked intrinsic signals in the retina. The retinas of five adult cats were examined in vivo. Animals were anesthetized and paralyzed for imaging stability. The retinas were imaged through a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service