Skip to Content
Merck
All Photos(3)

Documents

M9171

Sigma-Aldrich

D-(+)-Maltose monohydrate

≥99% (HPLC), BioXtra

Synonym(s):

4-O-α-D-Glucopyranosyl-D-glucose, Maltobiose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11 · H2O
CAS Number:
Molecular Weight:
360.31
Beilstein:
93798
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21

biological source

potato (tuber)

Quality Level

product line

BioXtra

Assay

≥99% (HPLC)

form

powder

technique(s)

HPLC: suitable

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter
≤0.3% glucose
≤1.0% maltotriose

ign. residue

≤0.1%

color

white

useful pH range

5.0-7 (25 °C, 180 g/L)

mp

125.13 °C (1013.25 hPa)

solubility

H2O: 1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.003%
sulfate (SO42-): ≤0.005%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

O.OC[C@@H](O)[C@@H](O[C@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C=O

InChI

1S/C12H22O11.H2O/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12;/h1,4-12,14-21H,2-3H2;1H2/t4-,5+,6+,7+,8+,9-,10+,11+,12+;/m0./s1

InChI key

HBDJFVFTHLOSDW-DNDLZOGFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

D-(+)-Maltose monohydrate has been used:
  • as a substrate for the detection of carbohydrase activity
  • as a carbohydrate supplement in yeast nitrogen base (YNB) medium for culturing the fungus Leucoagaricus gongylophorus
  • as substrate maltase enzyme activity of Leucoagaricus gongylophorus and to determine Michaellis-Menten constant (Km)

as a reference standard to investigate the mass spectra using gas chromatography-time of flight mass spectroscopy (GC-TOFMS)

Biochem/physiol Actions

Maltose is a disaccharide containing two glucose molecules with an α(1→4) glycosidic linkage. Maltose can be derived from starch in food through the action of amylase. Maltose can be found in many food products, including beer, cereals, and pasta.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fragmentation patterns of methyloxime-trimethylsilyl derivatives of constitutive mono-and disaccharide isomers analyzed by gas chromatography/field ionization mass spectrometry
Furuhashi T, et al.
Rapid Communications in Mass Spectrometry, 29(3), 238-246 (2015)
Starch metabolism in Leucoagaricus gongylophorus, the symbiotic fungus of leaf-cutting ants
Silva A, et al.
Microbiological research, 161(4), 299-303 (2006)
A metabolic pathway assembled by enzyme selection may support herbivory of leaf-cutter ants on plant starch
Bacci Jr M, et al.
Journal of Insect Physiology, 59(5), 525-531 (2013)
Hydrolytic enzymes of leaf-cutting ant fungi
Erthal Jr M, et al.
Comparative Biochemistry and Physiology. Part B, Biochemistry & Molecular Biology, 152(1), 54-59 (2009)
Jeshlee Vijayakumar et al.
Nature communications, 10(1), 2593-2593 (2019-06-15)
Prion-like domains (PLDs), defined by their low sequence complexity and intrinsic disorder, are present in hundreds of human proteins. Although gain-of-function mutations in the PLDs of neuronal RNA-binding proteins have been linked to neurodegenerative disease progression, the physiological role of

Protocols

Follow our procedure for the determination of alpha-Amylase activity. This enzymatic assay of a-Amylase guides you through the entire process and necessary calculations.

Follow our procedure for the determination of alpha-Amylase activity. This enzymatic assay of a-Amylase guides you through the entire process and necessary calculations.

Follow our procedure for the determination of alpha-Amylase activity. This enzymatic assay of a-Amylase guides you through the entire process and necessary calculations.

Follow our procedure for the determination of alpha-Amylase activity. This enzymatic assay of a-Amylase guides you through the entire process and necessary calculations.

Chromatograms

application for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service