Skip to Content
Merck
All Photos(1)

Key Documents

M1851000

Metronidazole benzoate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Methyl-5-nitro-1H-imidazole-1-ethyl benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13N3O4
CAS Number:
Molecular Weight:
275.26
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

metronidazole

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[N+](=O)([O-])c1[n](c(nc1)C)CCOC(=O)c2ccccc2

InChI

1S/C13H13N3O4/c1-10-14-9-12(16(18)19)15(10)7-8-20-13(17)11-5-3-2-4-6-11/h2-6,9H,7-8H2,1H3

InChI key

CUUCCLJJOWSASK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Metronidazole benzoate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[HPLC determination of benzoylmetronidazole and metronidazole in biological materials].
J Avramova
Die Pharmazie, 43(6), 436-436 (1988-06-01)
B Bloch et al.
South African medical journal = Suid-Afrikaanse tydskrif vir geneeskunde, 67(12), 455-457 (1985-03-23)
Metronidazole tablets, benzoyl metronidazole suspension and tinidazole tablets each given as a single 2 g bolus dose in an open controlled prospective study were found to be effective forms of treatment for Trichomonas vaginalis vaginitis. The cure rates on wet
G Takács
Therapia Hungarica (English edition), 40(4), 189-191 (1992-01-01)
Klion suspension was given to 42 girl and woman patients suffering from vaginal trichomoniasis. Negativity for Trichomonas was recorded in 73.8% of the cases in response to the first 10-day cure, and in 90.5% of the patients following repeated cures.
M A Homeida et al.
The Journal of antimicrobial chemotherapy, 18(2), 213-219 (1986-08-01)
A suspension of benzoylmetronidazole (6.4% w/v) was given orally at a dose of 15-25 ml, equivalent to 0.6-1 g metronidazole, once a day for three days to 11 children with giardiasis. Blood samples were collected after the first and third
A comparison of the pharmacokinetics of metronidazole in man after oral administration of single doses of benzoylmetronidazole and metronidazole.
G W Houghton et al.
British journal of clinical pharmacology, 14(2), 201-206 (1982-08-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service