Skip to Content
Merck
All Photos(1)

Key Documents

M1824990

Metoclopramide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-Amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide, Methoxychloroprocainamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22ClN3O2
CAS Number:
Molecular Weight:
299.80
Beilstein:
1884366
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

metoclopramide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCN(CC)CCNC(=O)c1cc(Cl)c(N)cc1OC

InChI

1S/C14H22ClN3O2/c1-4-18(5-2)7-6-17-14(19)10-8-11(15)12(16)9-13(10)20-3/h8-9H,4-7,16H2,1-3H3,(H,17,19)

InChI key

TTWJBBZEZQICBI-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Metoclopramide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D E Reed et al.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society, 26(11), 1663-1668 (2014-09-30)
Intestinal transit assessment in mice using existing methods requires long recording periods or euthanization of animals to localize a tracer. We have developed a novel in vivo method to assess gastrointestinal (GI) transit in mice based on a clinically used
B M Mishriky et al.
British journal of anaesthesia, 108(3), 374-383 (2012-02-07)
Nausea and vomiting occur commonly during and after Caesarean delivery (CD) performed under neuraxial anaesthesia. Metoclopramide is a prokinetic agent reported to be safe in parturients. This meta-analysis assesses the efficacy of metoclopramide for prophylaxis against intra- and postoperative nausea
Paul M Simpson et al.
Emergency medicine Australasia : EMA, 23(4), 452-457 (2011-08-10)
The objective of the present study was to conduct a systematic review and meta-analysis of randomized controlled trials, comparing metoclopramide with placebo, for preventing vomiting in patients who have received i.v. morphine for acute pain in the emergency setting, and
P Jay Pasricha et al.
Nature clinical practice. Gastroenterology & hepatology, 3(3), 138-148 (2006-03-03)
Metoclopramide, the only drug approved by the FDA for treatment of diabetic gastroparesis, but used off-label for a variety of other gastrointestinal indications, has many potentially troublesome adverse neurologic effects, particularly movement disorders. In this article, we comprehensively review the
Acute dystonic reaction caused by metoclopramide, versus tetanus.
Kyra Dingli et al.
BMJ (Clinical research ed.), 334(7599), 899-900 (2007-04-28)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service