Skip to Content
Merck
All Photos(1)

Documents

M1770200

Methyl stearate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Methyl octadecanoate, Stearic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16CO2CH3
CAS Number:
Molecular Weight:
298.50
Beilstein:
1786213
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

methyl stearate

manufacturer/tradename

EDQM

bp

181-182 °C/4 mmHg (lit.)

mp

37-41 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OC

InChI

1S/C19H38O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h3-18H2,1-2H3

InChI key

HPEUJPJOZXNMSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Methyl stearate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sonja Klee et al.
Rapid communications in mass spectrometry : RCM, 28(14), 1591-1600 (2014-06-05)
Atmospheric pressure chemical ionization (APCI) sources operated with point to plane DC discharges ('Coronas') frequently suffer from point electrode degradation and potentially lead to oxidation and/or fragmentation of the generated analyte ions. It is postulated that these adverse effects are
Oier Aizpurua-Olaizola et al.
Journal of food science, 80(1), E101-E107 (2014-12-05)
In this study, supercritical fluid extraction has been successfully applied to a sequential fractionation of fatty acids and polyphenols from wine wastes (2 different vitis vinifera grapes). To this aim, in a 1st step just fatty acids were extracted and
Leila Hejazi et al.
Rapid communications in mass spectrometry : RCM, 29(1), 91-99 (2014-12-03)
Mass spectrometric identification of compounds in chromatography can be obtained from molecular masses from soft ionization mass spectrometry techniques such as field ionization (FI) and fragmentation patterns from hard ionization techniques such as electron ionization (EI). Simultaneous detection by EI

Protocols

HPLC Analysis of Fatty Acid Methyl Esters (FAMES) on SUPELCOSIL™ LC-18

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service