Skip to Content
Merck
All Photos(1)

Key Documents

LIGHTOX58

Sigma-Aldrich

LightOx58

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H25NO2
CAS Number:
Molecular Weight:
383.48
UNSPSC Code:
12352101
NACRES:
NA.22

Assay

≥95% (HPLC)

form

powder

storage temp.

2-8°C

SMILES string

N1(CCC(c2c1ccc(c2)C#Cc3ccc(cc3)\C=C\C(=O)OC)(C)C)CC#C

InChI

1S/C26H25NO2/c1-5-17-27-18-16-26(2,3)23-19-22(12-14-24(23)27)11-10-20-6-8-21(9-7-20)13-15-25(28)29-4/h1,6-9,12-15,19H,16-18H2,2-4H3/b15-13+

InChI key

NLKZPAVDCYEIPQ-FYWRMAATSA-N

Application

Fluorescence microscopy, click chemistry, fluorescence spectroscopy.

Features and Benefits

LightOx58 is a fluorescent molecule that exhibits solvatochromatic fluorescence emission when excited with UV-A and violet (405 nm) light sources. In the free, unconjugated form LightOx58 can be used as an intracellular lipid droplet stain, and when photoactivated in a cellular environment, causes cell death through the release of reactive oxygen species (ROS). The N-propargyl group can also be utilized for ′click′ chemistry attachment to an azide-substituted molecule. In a non-polar cellular environment, peak emission wavelength is observed at 475 nm. Stock solutions are typically prepared at 10 mM in DMSO or 1 mM in EtOH.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Other Notes

Chemical Equivalency 1ea=1mg

Legal Information

LightOx is a trademark of LightOx Ltd.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Fluorescent retinoic acid analogues for use in cellular imaging, protein-ligand binding assays, and flow cytometry to bypass traditional problems associated with fluorescent probes.

Fluorescent retinoic acid analogues for use in cellular imaging, protein-ligand binding assays, and flow cytometry to bypass traditional problems associated with fluorescent probes.

Fluorescent retinoic acid analogues for use in cellular imaging, protein-ligand binding assays, and flow cytometry to bypass traditional problems associated with fluorescent probes.

Fluorescent retinoic acid analogues for use in cellular imaging, protein-ligand binding assays, and flow cytometry to bypass traditional problems associated with fluorescent probes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service