Skip to Content
Merck
All Photos(1)

Key Documents

L5783

Sigma-Aldrich

Lidocaine N-ethyl bromide

analytical standard, for drug analysis

Synonym(s):

N-(2,6-Dimethylphenylcarbamoylmethyl)triethylammonium bromide, QX-314

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H27BrN2O
CAS Number:
Molecular Weight:
343.30
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

[Br-].CC[N+](CC)(CC)CC(=O)Nc1c(C)cccc1C

InChI

1S/C16H26N2O.BrH/c1-6-18(7-2,8-3)12-15(19)17-16-13(4)10-9-11-14(16)5;/h9-11H,6-8,12H2,1-5H3;1H

InChI key

DLHMKHREUTXMCH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Lidocaine N-ethyl bromide has been used as a standard in studying the demethylation effect of lidocaine N-ethyl bromide on breast cancer cells in vitro using colorimetric assay.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

A quaternary derivative of lidocaine that is not membrane permeable. Blocks both fast Na+-dependent action potentials and voltage-dependent, non-inactivating Na+ conductance.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lidocaine and ropivacaine, but not bupivacaine, demethylate deoxyribonucleic acid in breast cancer cells in vitro
Lirk.P, et al.
British Journal of Anaesthesia, 113 (2014)
D P Roberson et al.
British journal of pharmacology, 164(1), 48-58 (2011-04-05)
We have developed a strategy to target the permanently charged lidocaine derivative lidocaine N-ethyl bromide (QX-314) selectively into nociceptive sensory neurons through the large-pore transient receptor potential cation channel subfamily V (TRPV1) noxious heat detector channel. This involves co-administration of
Kimbria J Blake et al.
Nature communications, 9(1), 37-37 (2018-01-04)
The hallmark of many bacterial infections is pain. The underlying mechanisms of pain during live pathogen invasion are not well understood. Here, we elucidate key molecular mechanisms of pain produced during live methicillin-resistant Staphylococcus aureus (MRSA) infection. We show that
Christian Brenneis et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(1), 315-326 (2013-01-04)
Specific somatosensations may be processed by different subsets of primary afferents. C-fibers expressing heat-sensitive TRPV1 channels are proposed, for example, to be heat but not mechanical pain detectors. To phenotype in rats the sensory function of TRPV1(+) afferents, we rapidly
Richard I Kuo et al.
Neuron, 73(5), 1016-1027 (2012-03-13)
Both human speech and animal vocal signals contain frequency-modulated (FM) sounds. Although central auditory neurons that selectively respond to the direction of frequency modulation are known, the synaptic mechanisms underlying the generation of direction selectivity (DS) remain elusive. Here we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service