Skip to Content
Merck

ICS024

Thiosulfate IC Standard

analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

grade

analytical standard

Quality Level

CofA

current certificate can be downloaded

manufacturer/tradename

RTC ICS-024-100

technique(s)

ion chromatography: suitable

application(s)

environmental
food and beverages

format

single component solution

storage temp.

2-30°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
ICS023ICS006ICS012
application(s)

environmental
food and beverages

application(s)

environmental
food and beverages

application(s)

environmental
food and beverages

application(s)

environmental
food and beverages

technique(s)

ion chromatography: suitable

technique(s)

ion chromatography: suitable

technique(s)

ion chromatography: suitable

technique(s)

ion chromatography: suitable

format

single component solution

format

single component solution

format

single component solution

format

single component solution

Quality Level

100, 300

Quality Level

300

Quality Level

300

Quality Level

100, 300

grade

analytical standard

grade

analytical standard, certified reference material

grade

certified reference material

grade

analytical standard

storage temp.

2-30°C

storage temp.

2-8°C

storage temp.

2-30°C

storage temp.

2-8°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Storage Class

10 - Combustible liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Global Trade Item Number

SKUGTIN
ICS024-100ML04061833865903

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service