Skip to Content
Merck
All Photos(1)

Key Documents

I0330000

Iopanoic acid

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-(3-Amino-2,4,6-triiodobenzyl)butyric acid, 3-(3-Amino-2,4,6-triiodophenyl)-2-ethylpropanoic acid, Iodopanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12I3NO2
CAS Number:
Molecular Weight:
570.93
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

iopanoic acid

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C(O)C(CC)CC1=C(I)C(N)=C(I)C=C1I

InChI

1S/C11H12I3NO2/c1-2-5(11(16)17)3-6-7(12)4-8(13)10(15)9(6)14/h4-5H,2-3,15H2,1H3,(H,16,17)

InChI key

OIRFJRBSRORBCM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Iopanoic acid EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Piero Berti et al.
Langenbeck's archives of surgery, 392(6), 709-713 (2006-11-15)
Amiodarone-induced thyrotoxicosis is a life-threatening condition. A prompt control of thyrotoxicosis is obtained by thyroidectomy. Preparation with iopanoic acid proved to be very effective in reducing cardiovascular complications. Nevertheless, general anesthesia and extensive surgery may affect negatively patients also after
Oral cholecystography with iopanoic acid.
R N Berk et al.
The New England journal of medicine, 290(4), 204-210 (1974-01-24)
Kostja Renko et al.
Endocrinology, 153(5), 2506-2513 (2012-03-22)
Enzymatic 5'- and 5-deiodination are key reactions for local and systemic activation and inactivation of iodothyronines and thyronamines. Expression of the three deiodinase (DIO) isoenzymes is regulated by a number of parameters, including thyroid status, genotype, micronutrient availability, and disease-related
Kurt Benkestock et al.
Rapid communications in mass spectrometry : RCM, 19(12), 1637-1643 (2005-05-26)
Most proteins in blood plasma bind ligands. Human serum albumin (HSA) is the main transport protein with a very high capacity for binding of endogenous and exogenous compounds in plasma. Many pharmacokinetic properties of a drug depend on the level
Fernando Lizcano et al.
Clinical and experimental pharmacology & physiology, 35(9), 1085-1090 (2008-05-29)
To investigate the effects of iopanoic acid (IA) and carbimazole on increased activity of the hypothalamic-pituitary-adrenal (HPA) axis in hyperthyroidism, we studied 14 women with hyperthyroidism caused by Graves' disease (n + 11) or toxic multinodular goitre (n + 3)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service