Skip to Content
Merck
All Photos(1)

Key Documents

I0150000

Indapamide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

N-(4-Chloro-3-sulfamoylbenzamido)-2-methylindoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16ClN3O3S
CAS Number:
Molecular Weight:
365.83
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

indapamide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC1Cc2ccccc2N1NC(=O)c3ccc(Cl)c(c3)S(N)(=O)=O

InChI

1S/C16H16ClN3O3S/c1-10-8-11-4-2-3-5-14(11)20(10)19-16(21)12-6-7-13(17)15(9-12)24(18,22)23/h2-7,9-10H,8H2,1H3,(H,19,21)(H2,18,22,23)

InChI key

NDDAHWYSQHTHNT-UHFFFAOYSA-N

Gene Information

human ... SLC12A3(6559)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Indapamide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Lact. - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Damien et al.
Clinical pharmacokinetics, 37 Suppl 1, 13-19 (1999-09-24)
In accordance with international guidelines recommending the use of low doses of antihypertensive agents, a new formulation of indapamide--indapamide sustained release (SR)--has been developed. Indapamide has been used worldwide for many years as an immediate release (IR) formulation at a
Bernard Waeber et al.
American journal of cardiovascular drugs : drugs, devices, and other interventions, 9(5), 283-291 (2009-10-02)
Patients with type 2 diabetes mellitus exhibit a marked increase in cardiovascular and renal risk. A number of interventional trials have shown that these patients benefit greatly from aggressive BP lowering, especially when the drug regimen comprises an inhibitor of
Luis M Ruilope et al.
Journal of hypertension. Supplement : official journal of the International Society of Hypertension, 24(3), S9-12 (2006-05-26)
Type 2 diabetes mellitus (T2DM) is often accompanied by high blood pressure (BP) and the clustering of several cardiovascular risk factors, and is the most frequent cause of end-stage renal disease. The stages of development of overt nephropathy in T2DM
G Leonetti
Drugs, 59 Suppl 2, 27-38 (2000-03-11)
Indapamide sustained release (SR) 1.5mg is a new galenic formulation that is characterised by a relatively constant plasma concentration at steady state, with only minor fluctuations during the 24-hour period. A dose-titration study of 3 doses of indapamide SR (1.5
Sheng Kang et al.
Clinical therapeutics, 26(2), 257-270 (2004-03-25)
A low-dose combination of perindopril and indapamide may effectively reduce blood pressure (BP) in hypertensive patients, but some factors related to study design might have contributed to the between-group differences in the rate of reduction of BP observed in some

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service